Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

481412

Sigma-Aldrich

NF-kB Activation Inhibitor IV

The NF-kB Activation Inhibitor IV, also referenced under CAS 139141-12-1, controls the biological activity of NF-kB. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

NF-kB Activation Inhibitor IV, (E)-2-Fluoro-4ʹ-methoxystilbene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13FO
CAS Number:
Molecular Weight:
228.26
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

DMSO: 20 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable trans-stilbene Resveratrol (Cat. No. 554325) analog that exhibits much enhanced anti-inflammatory potency, but no anti-oxidant activity. Suppresses LPS-induced COX-2 mRNA expression in BV-2 cells (~46% inhibition at 150 nM) and Shown to be ~130-fold more potent than Resveratrol in inhibiting TNF-α-stimulated NF-κB reporter activity in 293T cells (IC50 = 150 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
NF-κB Activation
Product does not compete with ATP.
Reversible: no
Target IC50: 150 nM inhibiting TNF-α-stimulated NF-κB reporter activity in 293T cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Heynekamp, J.J., et al. 2006. J. Med. Chem.49, 7182.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pascal Gelebart et al.
Translational oncology, 4(1), 9-19 (2011-02-03)
Mantle cell lymphoma (MCL) is a specific type of aggressive B-cell non-Hodgkin lymphoma. We recently found that IL-22RA1, one of the two subunits of the interleukin 22 (IL-22) receptor, is expressed in MCL cell lines but not benign lymphocytes. In
Je Sun Lim et al.
Cell death & disease, 13(11), 1002-1002 (2022-11-27)
Glioblastoma (GBM) is a highly vascular malignant brain tumor that overexpresses vascular endothelial growth factor (VEGF) and phosphofructokinase 1 platelet isoform (PFKP), which catalyzes a rate-limiting reaction in glycolysis. However, whether PFKP and VEGF are reciprocally regulated during GBM tumor
Yun-Ru Jaoying Huang et al.
Journal of cellular physiology, 237(12), 4517-4530 (2022-10-18)
Golgi apparatus (GA) and centrosome reposition toward cell leading end during directional cell migration in a coupling way, thereby determining cell polarity by transporting essential factors to the proximal plasma membrane. The study provides mechanistic insights into how GA repositioning

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service