Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

481001

Sigma-Aldrich

Anti-Neutrophil Elastase Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Neutrophil elastase antibody, Rabbit monoclonal antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ELANE(1991)

General description

Ammonium sulfate precipitated rabbit polyclonal antibody. Recognizes human neutrophil elastase.
Recognizes the neutrophil elastase protein.
This Anti-Neutrophil Elastase Rabbit pAb is validated for use in ELISA, Immunoblotting, Immunoelectrophoresis, Immunoprecipitation for the detection of Neutrophil Elastase.

Immunogen

Human
human neutrophil elastase

Application

ELISA (≥ 1:1000)

Immunoblotting (see comments)

Immunoelectrophoresis (see comments)

Immunoprecipitation (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Single arc by immunoelectrophoresis against neutrophil granule extract and human neutrophil elastase. This antibody has also been reported to work for immunoblotting and immunoprecipitation. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elaine F Kenny et al.
eLife, 6 (2017-06-03)
Neutrophils release neutrophil extracellular traps (NETs) which ensnare pathogens and have pathogenic functions in diverse diseases. We examined the NETosis pathways induced by five stimuli; PMA, the calcium ionophore A23187, nigericin, Candida albicans and Group B Streptococcus. We studied NET
Dae-goon Yoo et al.
Journal of immunology (Baltimore, Md. : 1950), 192(10), 4728-4738 (2014-04-18)
Cystic fibrosis (CF) airways are characterized by bacterial infections, excess mucus production, and robust neutrophil recruitment. The main CF airway pathogen is Pseudomonas aeruginosa. Neutrophils are not capable of clearing the infection. Neutrophil primary granule components, myeloperoxidase (MPO) and human
Avinash Khandagale et al.
Journal of leukocyte biology, 104(6), 1199-1213 (2018-08-15)
Mutations in the gene JAGN1 were recently discovered in patients with severe congenital neutropenia (SCN). Neutrophils release neutrophil extracellular traps (NETs) consisting of decondensed chromatin decorated with various granular proteins such as neutrophil elastase and myeloperoxidase (MPO) to combat microbial
Ava Hosseinzadeh et al.
Frontiers in immunology, 3, 391-391 (2012-12-28)
To prevent the spread of pathogens neutrophils as the first line of defense are able to release Neutrophil Extracellular Traps (NETs), a recently discovered form of immune response. Reactive oxygen species (ROS) have been shown to be essential for many
Kelly K Hunt et al.
Breast cancer research : BCR, 15(1), R3-R3 (2013-01-17)
Elafin is an elastase-specific inhibitor with increased transcription in normal mammary epithelial cells compared to mammary carcinoma cells. In this report, we test the hypothesis that inhibition of elastase, through induction of elafin, leads to inhibition of human breast cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service