Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C51H75N10O12
Molecular Weight:
1020.20
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light
Quality Segment
assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, desiccated (hygroscopic), protect from light
color
white
solubility
DMSO: 50 mg/mL
shipped in
wet ice
storage temp.
−20°C
General description
A cell-permeable, synthetic maleimido-conjugated VIVIT motif peptide (HPVIVIT) that acts as a potent bipartite inhibitor of Nuclear factor of activated T-cells (NFAT) (IC50 = 62 nM). Blocks NFAT-calcineurin interaction by targeting two separate calcineurin docking motifs. Completely blocks NFAT dephosphorylation at 1 mM. Determined to be more potent that Cyclosporine A (Cat. No. C3662) and VIVIT peptide (MAGPHPVIVITGPHEE) in blocking NFAT activity. Also shown to blocks NFAT-mediated T-cell activation and vascular smooth muscle cell proliferation, thereby reducing neoimtima (new thickening of arterial wall) formation in mouse models of restenosis (repeat blocking of blood vessels). The biological half life of MCV1 has been estimated to be about 36 hours in COS-1 cells.
Packaging
Packaged under inert gas
Physical form
Supplied as a trifluoroacetate salt.
Preparation Note
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Yu, H., et al. 2012. Circulation Res.110, 200.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.