Skip to Content
MilliporeSigma
All Photos(1)

Documents

476495

Sigma-Aldrich

PKD Inhibitor, CID755673

The PKD Inhibitor, CID755673, also referenced under CAS 521937-07-5, controls the biological activity of PKD. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PKD Inhibitor, CID755673, 7-Hydroxy-2,3,4,5-tetrahydro-1H-[1]benzofuro[2,3-c]azepin-1-one, PKCμ Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11NO3
CAS Number:
Molecular Weight:
217.22
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brown

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C12H11NO3/c14-7-3-4-10-9(6-7)8-2-1-5-13-12(15)11(8)16-10/h3-4,6,14H,1-2,5H2,(H,13,15)

InChI key

AACFPJSJOWQNBN-UHFFFAOYSA-N

General description

A cell-permeable benzoxoloazepinolone compound that effectively inhibits syntide-2 (Cat. No. 05-23-4910) phosphorylation by all three PKD/PKCµ isoforms 1, 2, & 3 (IC50 = 182, 280, and 227 nM, respectively), while exhibiting ≥30-fold less potency towards Cdk activating kinase, Plk1, CaMKIIα, as well as 44 other kinases, 33 of which are not significantly inhibited even at concentrations as high as 10 µM, including Akt1/2 and PKCα/β/δ/ζ. Shown to block PMA- (Cat. No. 524400) induced PKD1 phosphorylation on Ser742 and Ser916 in LNCaP cells, PMA-induced HCAS5 nuclear export in HeLa cells, as well as other PKD-mediated cellular responses. The mode of inhibition is demonstrated not to be ATP-competitive in nature. Also reported to inhibit MAPKAPK2, GSK-3β, CKIδ, MK5 (PRAK), CDK2, and Erk1 kinase activity at much elevated concentrations (% inhibition by 10 µM inhibitor = 95, 86, 82, 75, 71, and 50, respectively).
A cell-permeable benzoxoloazepinolone compound that effectively inhibits syntide-2 (Cat. No. 05-23-4910) phosphorylation by all three PKD/PKCµ isoforms 1, 2, & 3 (IC50 = 182, 280, and 227 nM, respectively), while exhibiting ≥30-fold less potency towards Cdk activating kinase, Plk1, CaMKIIα, as well as 44 other kinases, 33 of which are not significantly inhibited even at concentrations as high as 10 µM, including Akt1/2 and PKCα/β/δ/ζ. Shown to block PMA- (Cat. No. 524400) induced PKD1 phosphorylation on Ser742 and Ser916 in LNCaP cells, PMA-induced HCAS5 nuclear export in HeLa cells, as well as other PKD-mediated cellular responses. The mode of inhibition is demonstrated not to be ATP-competitive in nature. Also reported to inhibit MAPKAPK2, GSK-3β, CKIδ, MK5 (PRAK), CDK2, and Erk1 kinase activity at much elevated concentrations (% inhibition by 10 µM inhibitor = 95, 86, 82, 75, 71, and 50, respectively).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

LaValle, C.R., et al. 2010. BMC Chemical Biology10, 5.
Sharlow, E.R., et al. 2008. J. Biol. Chem.283, 33516.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph H Lucas et al.
Toxicological sciences : an official journal of the Society of Toxicology, 190(2), 215-226 (2022-09-16)
Perfluorooctane sulfonic acid (PFOS) is a long chain per- and polyfluoroalklyl substance (PFAS) that has been used in aqueous film-forming foams. Emerging epidemiological evidence indicates that PFOS may be associated with chronic lung diseases such as asthma and analysis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service