Skip to Content
MilliporeSigma

475889

MK-886

≥99% (TLC), COX pathway blocker, solid

Synonym(s):

MK-886, 3-[1-( p-Chlorobenzyl)-5-(isopropyl)-3- t-butylthioindol-2-yl]-2,2-dimethylpropanoic Acid, Na, COX-1 Inhibitor III, FLAP Inhibitor I, MK886, mPGES-1 Inhibitor I

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H33ClNO2S · Na
CAS Number:
Molecular Weight:
494.06
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥99% (TLC)
Form:
solid
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

MK-886, A cell-permeable, orally active NSAID that blocks cellular Cox pathway PGE2 production by inhibiting COX-1 and mPGES-1, but not COX-2, activity, as well as suppresses cellular 5-LO pathway activation by inhibiting FLAP, rather than 5-LO, activity.

Quality Segment

assay

≥99% (TLC)

form

solid

potency

102 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 25 mg/mL, ethanol: 25 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

S(CCCC)c1c2c([n](c1CC(C)(C)C(=O)O)Cc3ccc(cc3)Cl)ccc(c2)C(C)C

InChI

1S/C27H34ClNO2S/c1-6-7-14-32-25-22-15-20(18(2)3)10-13-23(22)29(17-19-8-11-21(28)12-9-19)24(25)16-27(4,5)26(30)31/h8-13,15,18H,6-7,14,16-17H2,1-5H3,(H,30,31)

InChI key

VFMGWQLOCZBFCK-UHFFFAOYSA-N

General description

A cell-permeable, orally active NSAID (nonsteroidal antiinflammatory drug) that blocks cellular Cox pathway PGE2 (prostaglandin E2) production by inhibiting COX-1 and mPGES-1 (microsomal PGE2 synthase-1), but not COX-2, activity (IC50 = 8, 2, and 58 M, respectively), as well as suppresses cellular 5-LO (5-Lypoxygenase; Cat. No. 437996) pathway activation by inhibiting FLAP (5-LO-activating protein), rather than 5-LO, activity (<10% by 1 M MK-886). Unlike NSAIDs (nonsteroidal antiinflammatory drugs) that target only COX pathway, MK-886 does not cause gastrointestinal damages when applied in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
leukotreine biosybthesis
Product does not compete with ATP.
Reversible: no

Other Notes

Koeberle, A., et al. 2009. Eur. J. Pharmacol.608, 84.
Koeberle, A., et al. 2008. J. Pharmacol. Exp. Ther.326, 975.
Fisher, L., et al. 2007. Br. J. Pharmacol.152, 471.
Ford-Hutchinson, A.W., et al. 1993. Can. J. Physiol. Pharmacol. 71, 806.
Ford-Hutchinson, A.W. 1991. Trends Pharmacol.12, 68.
Dixon, R.A., et al. 1990. Nature 343, 282.
Rouzer, C.A., et al. 1990. J. Biol. Chem.265, 1436.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library