Skip to Content
MilliporeSigma

443713

Maltotriose

A substrate for amylomaltase.

Synonym(s):

Maltotriose, Glcα1,4Glcα1,4Glc

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H32O16
CAS Number:
Molecular Weight:
504.44
UNSPSC Code:
12352201
NACRES:
NA.79
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: soluble

shipped in

ambient

storage temp.

15-25°C

SMILES string

O1C(C(C(C(C1CO)OC3OC(C(C(C3O)O)O)CO)O)O)OC2C(OC(C(C2O)O)O)CO

InChI

1S/C18H32O16/c19-1-4-7(22)8(23)12(27)17(31-4)34-15-6(3-21)32-18(13(28)10(15)25)33-14-5(2-20)30-16(29)11(26)9(14)24/h4-29H,1-3H2

InChI key

FYGDTMLNYKFZSV-UHFFFAOYSA-N

General description

A substrate for amylomaltase.

Application


  • Screening of α-amylase/trypsin inhibitor activity in wheat, spelt and einkorn by high-performance thin-layer chromatography.: The research investigates the inhibitory activity of α-amylase and trypsin in different wheat species using high-performance thin-layer chromatography. Maltotriose is utilized as a substrate to evaluate enzyme activity, contributing to understanding enzyme inhibition in cereals (Müller et al., 2024).

Other Notes

Terada, Y., et al. 1999. Appl. Environ. Microbiol. 65, 910.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under Maltotriose


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library