Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

440153

Sigma-Aldrich

L-α-lysophosphatidylinositol sodium salt

>99% (HPLC), powder, GPR55 agonist, Calbiochem®

Synonym(s):

L-α-lysophosphatidylinositol Sodium Salt, Soy, L-α-lysophosphatidylinositol LPI, Lyso PI, Soy Lyso PI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H48NaO12P
CAS Number:
Molecular Weight:
594.61
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

L-α-lysophosphatidylinositol Sodium Salt, Soy, A potent agonist and assumed endogenous ligand of GPR55.

Quality Level

Assay

>99% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

O[C@H]1[C@H](OP(OC[C@H](O)COC(CCCCCCCCCCCCCCC)=O)([O-][Na+])=O)[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

General description

A potent agonist and assumed endogenous ligand of GPR55, shown to induce rapid phosphorylation of p38 mitogen-activated protein kinase (MAPK) and its downstream activating transcription factor 2 (ATF2) in both GPR55-transfected HEK293 cells, and in IM-9 lymphoblastoid cells naturally expressing GPR55 at 1 µM, while having no effect on vector-transfected cells. In addition, shown to enhance serum induced cell migration in MCF-7 cells over expressing GPR55.
A potent agonist and assumed endogenous ligand of GPR55, shown to induce rapid phosphorylation of p38 mitogen-activated protein kinase (MAPK) and its downstream activating transcription factor 2 (ATF2) in both GPR55-transfected HEK293 cells, and in IM-9 lymphoblastoid cells naturally expressing GPR55 at 1uM, while having no effect on vector-transfected cells. In addition, shown to enhance serum induced cell migration in MCF-7 cells over expressing GPR55.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Oka, S., et al. 2010. J Biochem.147, 671.
Oka, S., et al. 2007.
Biochem. Biophys. Res. Commun.362, 928.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service