Skip to Content
MilliporeSigma

437650

Lipopolysaccharide

from Salmonella typhimurium, , lyophilized, iNOS stimulator, Calbiochem

Synonym(s):

Salmonella LPS, Salmonella endotoxin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352201
NACRES:
NA.77
Form:
lyophilized
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Lipopolysaccharide, Salmonella typhimurium, Stimulates the activity of inducible nitric oxide synthase.

description

RTECS - OJ0895637

Quality Segment

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤2% nucleic acid, ≤2% protein

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

Endotoxin lipopolysaccharides initiate a variety of biochemical cell activation pathways (PKC, cAMP-dependent protein kinase, arachidonate metabolism, protein myristoylation, and activation of G-proteins). These unique macromolecules have been extensively studied in order to elucidate and define revelent pathophysiological parameters of endotoxin shock, a profound life-threatening consequence of bacterial sepsis. More recently, these bacterial products have generated intense interest due to their pluripotential immunostimulatory activity, as manifested by the activation of host cells (e.g. B lymphocytes, macrophages) to functional differentiation. Host cells activated by lipopolysaccharides produce a spectrum of hormone-active lymphokines and monokines, including interferons (α, β, and γ), interleukins (IL-1 and IL-6), tumor necrosis factor, platelet activating factor and procoagulant tissue factor. Stimulates the activity of inducible nitric oxide synthase. Induces apoptosis in mouse thymus and swine lymphocytes.
Stimulates the activity of inducible nitric oxide synthase. Induces apoptosis in mouse thymus and swine lymphocytes.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Inducible nitric oxide synthase
Product does not compete with ATP.
Reversible: no

Preparation Note

Following reconstitution, store in the refrigerator (4°C). Aqueous stock solutions are stable for up to 6 months at 4°C.

Other Notes

Hattori, S., et al. 1995. Biochem. Mol. Biol. Int. 35, 177.
Mayeux, P.R., et al. 1995. Biochem. Pharmacol. 49, 115.
Norimatsu, M., et al. 1995. Infect. Immun.63, 1122.
Feinstein, D.L., et al. 1993. J. Neurochem. 60, 1945.
Zhang, Y.H., et al. 1993. Infect. Immun.61, 5044.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful (C)


Still not finding the right product?


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library