Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

428015

Millipore

Sodium dodecyl sulfate

>97% (titration)

Synonym(s):

Sodium n-Dodecyl Sulfate, Sodium Lauryl Sulfate, SDS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H25O4S · Na
CAS Number:
Molecular Weight:
288.38
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.21

Product Name

Sodium dodecyl sulfate, An ionic detergent useful in electrophoretic separation of proteins and lipids.

Quality Level

Assay

>97% (titration)

form

solid

mol wt

micellar wt 18,000

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

aggregation number

62

color

white

CMC

7-10 mM

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Na+].[S](=O)(=O)([O-])OCCCCCCCCCCCC

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

General description

An ionic detergent useful in electrophoretic separation of proteins and lipids (aggregation number: 62; CMC: 7-10 mM; micellar weight: 18,000).

Application


  • Construction of dental pulp decellularized matrix by cyclic lavation combined with mechanical stirring and its proteomic analysis.: This study details the use of sodium dodecyl sulfate (SDS) in creating a dental pulp decellularized matrix through cyclic lavation and mechanical stirring, followed by proteomic analysis to identify the resulting protein composition. (Zhang et al., 2024).

  • Polycarbonate Ultracentrifuge Tube Re-use in Proteomic Analyses of Extracellular Vesicles.: The research explores the reusability of polycarbonate ultracentrifuge tubes in proteomic analyses of extracellular vesicles, utilizing SDS for efficient protein extraction and quantification. (Cahalane et al., 2024).

  • Identification of protein components of the transformation system in the cell line of immortalized human keratinocytes HaCaT exposed to surfactants.: This study identifies protein components in HaCaT cells exposed to surfactants, including SDS, providing insights into cellular transformations induced by these chemicals. (Shkrigunov et al., 2024).

  • Assessing the precision of a detergent-assisted cartridge precipitation workflow for non-targeted quantitative proteomics.: The research assesses the precision of a detergent-assisted precipitation workflow using SDS for non-targeted quantitative proteomics, demonstrating its utility in protein sample preparation. (Nickerson et al., 2024).

Warning

Toxicity: Harmful (C)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service