Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

420400

Sigma-Aldrich

Anti-KDEL Mouse mAb (10C3)

liquid, clone 10C3, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

10C3, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

mouse, sheep, canine, hamster, Drosophila, rat, guinea pig, human, monkey, yeast, rabbit, porcine

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Kdelr1(68137)

General description

Protein G purified mouse monoclonal antibody. Recognizes the KDEL Grp78 (BiP) and Grp94 proteins.
Recognizes the Grp78 (BiP) and Grp94 proteins.
This Anti-KDEL Mouse mAb (10C3) is validated for use in Immunoblotting, Immunocytochemistry, Immunoprecipitation for the detection of KDEL.

Immunogen

Rat
a synthetic peptide (SEKDEL) corresponding to amino acids 649-654 of rat Grp78, conjugated to KLH

Application

Immunoblotting (4 µg/ml, colorimetric)

Immunocytochemistry (5 µg/ml)

Immunoprecipitation (5 µg/ml)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Highly Toxic (H)

Physical form

In PBS, 50% glycerol.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
RK-13 cells, Mouse liver microsomes, Rat liver microsomes, Heat shocked HeLa cells

Other Notes

Also recognizes unidentified proteins at ~55 kDa and ~43 kDa by immunoblotting. Does not cross-react with Hsp70. Variables associated with assay conditions will dictate the proper working dilution.
Kuznetsov, G., et al. 1994. J. Biol. Chem.269, 22990.
Huovila, A.-P., et al. 1992. J. Cell Biol. 118, 1305.
Munro, S., and Pelham, H.R.B. 1987. Cell48, 899.
Munro, S., and Pelham, H.R.B. 1986. Cell46, 291.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vangelis Kondylis et al.
Autophagy, 9(6), 861-880 (2013-03-14)
Activation of TLR signaling has been shown to induce autophagy in antigen-presenting cells (APCs). Using high-resolution microscopy approaches, we show that in LPS-stimulated dendritic cells (DCs), autophagosomes emerge from MHC class II compartments (MIICs) and harbor both the molecular machinery
Smriti A Agrawal et al.
Human molecular genetics, 26(14), 2667-2677 (2017-05-06)
Retinitis pigmentosa (RP) is the most common form of inherited retinal dystrophy. We recently identified mutations in REEP6, which encodes the receptor expression enhancing protein 6, in several families with autosomal recessive RP. REEP6 is related to the REEP and
Michael A Robichaux et al.
Disease models & mechanisms, 15(5) (2022-03-12)
The P23H mutation in rhodopsin (Rho), the rod visual pigment, is the most common allele associated with autosomal-dominant retinitis pigmentosa (adRP). The fate of misfolded mutant Rho in rod photoreceptors has yet to be elucidated. We generated a new mouse
Gary P Brennan et al.
PloS one, 8(1), e54491-e54491 (2013-01-30)
14-3-3 proteins are ubiquitous molecular chaperones that are abundantly expressed in the brain where they regulate cell functions including metabolism, the cell cycle and apoptosis. Brain levels of several 14-3-3 isoforms are altered in diseases of the nervous system, including
Hyojeong Hwang et al.
Cell reports, 41(11), 111802-111802 (2022-12-15)
Asymmetric localization of mRNAs is crucial for cell polarity and cell fate determination. By performing fractionation RNA-seq, we report here that a large number of maternal RNAs are associated with the ER in Xenopus oocytes but are released into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service