Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

420104

Sigma-Aldrich

JAK3 Inhibitor II

The JAK3 Inhibitor II, also referenced under CAS 211555-04-3, controls the biological activity of JAK3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JAK3 Inhibitor II, 4-[(3ʹ-Bromo-4ʹ-hydroxyphenyl)amino]-6,7-dimethoxyquinazoline, WHI-P154

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14BrN3O3
CAS Number:
Molecular Weight:
376.20
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Brc1c(ccc(c1)Nc2ncnc3c2cc(c(c3)OC)OC)O

InChI

1S/C16H14BrN3O3/c1-22-14-6-10-12(7-15(14)23-2)18-8-19-16(10)20-9-3-4-13(21)11(17)5-9/h3-8,21H,1-2H3,(H,18,19,20)

InChI key

CBIAKDAYHRWZCU-UHFFFAOYSA-N

General description

A potent, cell-permeable, reversible, ATP-competitive, and specific inhibitor of JAK3 (IC50 = 5.6 µM). Has no effect on either JAK1 or JAK2. Has also been shown to prevent the ionizing radiation-induced activation of c-Jun in DT-40 cells. Also acts as a potent inhibitor of glioblastoma cell adhesion and migration.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
JAK-3
Product competes with ATP.
Reversible: yes
Target IC50: 5.6 µM against JAK3

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ghosh, S., et al. 2001. Acta Crystallogr. C.57, 76.
Sudbeck, E.A., et al. 1999. Clin. Cancer Res. 5, 1569.
Goodman, P.A., et al. 1998. J. Biol. Chem. 273, 17742.
Narla, R.K., et al. 1998. Clin Cancer Res.4, 2463.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E A Sudbeck et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 5(6), 1569-1582 (1999-07-02)
A novel homology model of the kinase domain of Janus kinase (JAK) 3 was used for the structure-based design of dimethoxyquinazoline compounds with potent and specific inhibitory activity against JAK3. The active site of JAK3 in this homology model measures
S Ghosh et al.
Acta crystallographica. Section C, Crystal structure communications, 57(Pt 1), 76-78 (2001-02-15)
The title compounds, C16H15BrN3(O3)(+).Cl(-).CH4O (WHI-P154) and C16H16N3(O3)(+).Cl(-) (WHI-P180), are potent inhibitors [WHI-P154 with IC50 = 5.6 microM and WHI-P180 with IC50 = 4.0 microM for epidermal growth factor receptor (EGFR) kinase inhibition] of the EGFR tyrosine kinase as well as
R K Narla et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 4(10), 2463-2471 (1998-10-31)
Glioblastoma multiforme is a highly invasive primary brain tumor with a disappointingly high local recurrence rate and mortality despite intensive multimodality treatment programs. Therefore, new agents that are capable of inhibiting the infiltration of normal brain parenchyma by glioblastoma cells
P A Goodman et al.
The Journal of biological chemistry, 273(28), 17742-17748 (1998-07-04)
Exposure of B-lineage lymphoid cells to ionizing radiation induces an elevation of c-jun proto-oncogene mRNA levels. This signal is abrogated by protein-tyrosine kinase (PTK) inhibitors, indicating that activation of an as yet unidentified PTK is mandatory for radiation-induced c-jun expression.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service