Skip to Content
MilliporeSigma

420101

JAK3 Inhibitor I

The JAK3 Inhibitor I, also referenced under CAS 202475-60-3, controls the biological activity of JAK3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JAK3 Inhibitor I, 4-(4ʹ-Hydroxyphenyl)amino-6,7-dimethoxyquinazoline, WHI-P131

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H15N3O3
CAS Number:
Molecular Weight:
297.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥97% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white to off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N(c3ccc(cc3)O)c1ncnc2c1cc(c(c2)OC)OC

InChI

1S/C16H15N3O3/c1-21-14-7-12-13(8-15(14)22-2)17-9-18-16(12)19-10-3-5-11(20)6-4-10/h3-9,20H,1-2H3,(H,17,18,19)

InChI key

HOZUXBLMYUPGPZ-UHFFFAOYSA-N

General description

A cell-permeable, reversible, potent, ATP-competitive, and specific inhibitor of JAK3 (Janus family kinase 3; IC50 = 78 µM). Has no effect on JAK1, JAK2, or Zap/Syk or Src tyrosine kinases. Acts as a potent inhibitor of glioblastoma cell adhesion and migration. Also reported to block thrombin-induced platelet aggregation.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Janus family kinase 3
Product competes with ATP.
Reversible: yes
Target IC50: 78 µM against Janus family kinase 3

Packaging

Packaged under inert gas

Other Notes

Tibbles, H.E., et al. 2001. J. Biol. Chem.276, 17815.
Sudbeck, E.A., et al. 1999. Clin. Cancer Res.5, 1569.
Goodman, P.A., et al. 1998. J. Biol. Chem. 273, 17742.
Narla, R.K., et al. 1998. Clin. Cancer Res. 4, 2463.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library