Recommended Products
Assay
≥98% (SDS-PAGE)
Quality Level
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
shipped in
ambient
storage temp.
−20°C
General description
Native IgG from rabbit plasma. IgG is a glycoprotein found in serum and tissue fluids. For use as a negative control in parallel with specific rabbit IgG-type primary antibodies.
Warning
Toxicity: Standard Handling (A)
Physical form
In 10 mM sodium phosphate, 0.85% NaCl, 0.05% NaN₃, pH 7.2.
Reconstitution
Following initial thaw, aliquot and freeze (-20°C). Avoid freeze/thaw cycles.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Gregory J Hogan et al.
PLoS biology, 13(11), e1002307-e1002307 (2015-11-21)
Reprogramming of a gene's expression pattern by acquisition and loss of sequences recognized by specific regulatory RNA binding proteins may be a major mechanism in the evolution of biological regulatory programs. We identified that RNA targets of Puf3 orthologs have
Jun Li et al.
Theranostics, 11(12), 5970-5985 (2021-04-27)
Overactivation of N-methyl-D-aspartate receptor (NMDAR) in the spinal cord dorsal horn (SDH) in the setting of injury represents a key mechanism of neuropathic pain. However, directly blocking NMDAR or its downstream signaling, interaction between postsynaptic density-95 (PSD-95) and neuronal nitric
Dasari Vasanthi et al.
Development (Cambridge, England), 137(24), 4239-4247 (2010-11-26)
Hox genes are necessary for proper morphogenesis and organization of various body structures along the anterior-posterior body axis. These genes exist in clusters and their expression pattern follows spatial and temporal co-linearity with respect to their genomic organization. This colinearity
Dasari Vasanthi et al.
Scientific reports, 3, 3011-3011 (2013-10-23)
Anterior-posterior body axis in all bilaterians is determined by the Hox gene clusters that are activated in a spatio-temporal order. This expression pattern of Hox genes is established and maintained by regulatory mechanisms that involve higher order chromatin structure and
Shama Bhatia et al.
Methods in molecular biology (Clifton, N.J.), 1965, 389-403 (2019-05-10)
Chromatin immunoprecipitation (ChIP) is widely used to measure protein-DNA interactions. This protocol outlines a ChIP method used to identify the association of a protein or protein modification (such as a specific histone modification-methylation, acetylation, etc.) of interest with a specific
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service