Skip to Content
MilliporeSigma

Skip To

4010-OP

Ethylenediaminetetraacetic acid disodium salt dihydrate

OmniPur® Grade, >= 99.0%

Synonym(s):

Ethylenediaminetetraacetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid, Disodium Salt, Dihydrate, (Ethylenedinitrilo)tetraacetic acid, Disodium Salt, Dihydrate, Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
UNSPSC Code:
12352107
NACRES:
NA.21
Beilstein/REAXYS Number:
3900609
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

suitable for SM 4500 - NH3

Quality Segment

assay

≥99.0%

form

crystalline powder

reaction suitability

reagent type: chelator

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

mp

248 °C (dec.) (lit.)

foreign activity

DNAse, none detected, Protease, none detected, RNAse, none detected

shipped in

ambient

SMILES string

O.O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

InChI key

OVBJJZOQPCKUOR-UHFFFAOYSA-L

General description

Molecular Biology Grade.

A260 (0.1 M, water): ≤0.2 AU
A280 (0.1 M, water): ≤0.02 AU
Assay: ≥99.0%
DNAse: none detected
Heavy metals (as Pb): ≤0.0005%
Loss on drying: ≤10.0%
Phosphatase: none detected
Protease: none detected
RNAse: none detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application

Chelator of divalent cations. Inhibits enzymes, such as metalloproteases, that require divalent cations for activity.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful (C)


Still not finding the right product?


Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Inhalation - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters