Skip to Content
MilliporeSigma

401010

I-BET

A cell-permeable benzodiazepine compound that binds the tandem bromodomains of BET (bromodomain and extra terminal domain) family members BRD2 (1-473), BRD3 (1-434), and BRD4 (1-477) with high affinity.

Synonym(s):

I-BET, (2-[(4chlorophenyl)-1-methyl-8-(methyloxy)-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepin-4-yl]-N-ethylacetamide, GSK525762A, (S)-2-(6-(4-Chlorophenyl)-8-methoxy-1-methyl-4H-benzo[f][1,2,4]triazolo[4,3-a][1,4]diazepin-4-yl)-N-ethylacetamide, BRD2 Inhi, GSK525762A, (S)-2-(6-(4-Chlorophenyl)-8-methoxy-1-methyl-4H-benzo[f][1,2,4]triazolo[4,3-a][1,4]diazepin-4-yl)-N-ethylacetamide, (2-[(4chlorophenyl)-1-methyl-8-(methyloxy)-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepin-4-yl]-N-ethylacetamide, BRD2 Inhibitor I, BRD3 Inhibitor I, BRD4 Inhibitor I

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H22ClN5O2
CAS Number:
Molecular Weight:
423.90
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Quality level:
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

yellow-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Clc1ccc(cc1)C2=N[C@H](c3[n](c(nn3)C)c4c2cc(cc4)OC)CC(=O)NCC

InChI

1S/C22H22ClN5O2/c1-4-24-20(29)12-18-22-27-26-13(2)28(22)19-10-9-16(30-3)11-17(19)21(25-18)14-5-7-15(23)8-6-14/h5-11,18H,4,12H2,1-3H3,(H,24,29)/t18-/m0/s1

InChI key

AAAQFGUYHFJNHI-SFHVURJKSA-N

General description

A cell-permeable benzodiazepine compound that binds the tandem bromodomains of BET (bromodomain and extra terminal domain) family members BRD2 (1-473), BRD3 (1-434), and BRD4 (1-477) with high affinity (Kd = 61.3, 50.5, and 55.2 nM, respectively, by ITC) and effectively competes against tetra-acetylated H4 peptide (Millipore cat. no. 12-379) for BRD2/3/4 binding (IC50 = 32.5, 42,4, and 36.1 nM, respectively, in competitive equilibrium binding assays), while exhibiting little affinity toward 5 other bromodomain-containing proteins (ATAD2, BAZ2B, CREBBP, PCAF, SP140) or a panel of 38 cellular enzymes, GPCRs, transporters, and ion channels. Shown to differentially modulate LPS-induced gene expression, notably suppressing LPS-induced upregulation of genes involved in inflammatory response, in murine BMDMs (bone marrow-derived macrophages) in vitro (30 min 1 µM pretreatment) and effectively prevent LPS-, heat-killed Salmonella typhimurium-, and CLP- (caecal ligation and puncture) induced death in mice in vivo (30 mg/kg i.v.). Cellular Brd2/3/4 knockdown using siRNA results in mostly the same effect as I-BET treatment in modulating LPS gene induction profile in murine BMDMs, but not without exceptions, indicating that not all BET-dependent transcription regulations are mediated via its binding to acetylated histones. Also available as a 50 mM solution in DMSO (Cat. No. 5.06071).

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Nicodeme, E., et al. 2010. Nature468, 1119.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Regulatory Review (Z)


Still not finding the right product?

Explore all of our products under I-BET


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



William R Shadrick et al.
Bioorganic & medicinal chemistry, 26(1), 25-36 (2017-11-25)
Within the last decade, the Bromodomain and Extra-Terminal domain family (BET) of proteins have emerged as promising drug targets in diverse clinical indications including oncology, auto-immune disease, heart failure, and male contraception. The BET family consists of four isoforms (BRD2
Julie A Johannessen et al.
iScience, 26(10), 107726-107726 (2023-09-18)
MLL-rearranged (MLL-r) leukemias are among the leukemic subtypes with poorest survival, and treatment options have barely improved over the last decades. Despite increasing molecular understanding of the mechanisms behind these hematopoietic malignancies, this knowledge has had poor translation into the



Global Trade Item Number

SKUGTIN
401010-5MG04055977189391