Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

400051

Sigma-Aldrich

Hygromycin B

from Streptomyces sp., >85% (HPLC), liquid, aminoglycoside antibiotic, Calbiochem®

Synonym(s):

Hygromycin B, Streptomyces sp.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H37N3O13
CAS Number:
Molecular Weight:
527.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Hygromycin B, Streptomyces sp., Unique aminoglycoside antibiotic that inhibits the growth of prokaryotic (bacteria) and eukaryotic microorganisms (yeasts) and mammalian cells.

Quality Level

Assay

>85% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

amber to brown

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C20H37N3O13/c1-23-7-2-5(21)9(26)15(10(7)27)33-19-17-16(11(28)8(4-25)32-19)35-20(36-17)18(31)13(30)12(29)14(34-20)6(22)3-24/h5-19,23-31H,2-4,21-22H2,1H3/t5-,6+,7+,8-,9+,10-,11+,12-,13-,14-,15-,16+,17+,18-,19+,20?/m1/s1

InChI key

GRRNUXAQVGOGFE-KPBUCVLVSA-N

General description

Bioassay: 350-450 units/mg Hygromycin B. 106 units is equivalent to ~900 mg.
Hygromycin B, an aminoglycoside antibiotic, inhibits the growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Specifically, it inhibits protein synthesis by interfering with translocation of the 70S ribosome and inducing misreading of the mRNA template (Dean, N., Gonzalez, A., et al., Rao, S.N., et al.). Hygromycin B has been used to select mutants in a wide variety of cells including bacteria (Bilang, R., et al., Salauze, D., et al.), protozoans (Lee, M.G-S., and Van der Ploeg, L.H.T.), yeast (Perlin, D.S., et al.), fungi (Cullen, D., et al., Kronstad, J.W., et al., Egelhoff, T.T., et al., Leslie, J.F., and Dickman, M.B., Bulte, L., and Bennoun, P.), plants (Dean, N., Damm, B., et al., Rikkerink, E. H., et al., Sugimoto, K., et al.), and mammalian cells (Crespi, C.L., et al., Giordano, T.J., and McAllister, W.T., Hubbard, S. C., et al.).



Resistance to Hygromycin B is conferred by a gene coding for a phosphotransferase that phosphorylates Hygromycin B, thereby inactivating it (Bilang, R., et al., Malpartida, F., et al.). Hygromycin B is known to selectively penetrate cells that have been rendered permeable by virus infection. This, combined with its potency in inhibiting translation, makes it an effective antiviral agent (MacIntyre, G., et al., Zhou, J., et al.).



The analytical data listed below will vary from lot to lot.



Bioassay: 350-450 U/mg

Concentration: 444,600 U/ml; 394 mg Hygromycin B/ml

Density: 1140-1155 mg/ml

% solids: 34.6%

Fill volumes:

100,000 U = 225 µl

1,000,000 U = 2.25 ml

10,000,000 U = 22.5 ml

250,000 U = 562 µl

5,000,000 U = 11.25 ml
Supplied with a directional insert. Note: 1 KU = 1000 units; 1 MU = 1,000,000 units.
Unique aminoglycoside antibiotic that inhibits the growth of prokaryotic (bacteria) and eukaryotic microorganisms (yeasts) and mammalian cells. Inhibits protein synthesis at the translocation step on the 70S ribosome and causes misreading of the mRNA. Hph, a gene from E. coli, encodes resistance to hygromycin B and can be isolated and cloned by recombinant DNA techniques. This hygromycin B-resistance gene is particularly useful for identification or selection of recombinant clones in a variety of cell types. Hygromycin B penetrates cells that have been permeabilized by virus infection and can act as an effective antiviral agent.

Biochem/physiol Actions

Primary Target
growth of prokaryotic (bacteria) and eukaryotic microorganisms (yeasts) and mammalian cells
Secondary Target
protein synthesis at the translocation step on the 70S ribosome and causes misreading of the mRNA
The activity of Hygromycin B is measured in a zonal inhibition assay using Bacillus subtilis. The activity is measured relative to a reference standard.

Warning

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Reconstitution

Aqueous stock solutions should be kept at concentrations of ≥50 mg/ml and can be stored at either 4°C or -20°C. Aqueous stock solutions (≥50 mg/ml) are stable for up to 6 months at 4°C or -20°C. Working solutions (<2 mg/ml) are stable for up to 1 month at 4°C.



Note: This product is highly toxic. Read enclosed MSDS before handling.

Other Notes

Dean, N. 1995. Proc. Natl. Acad. Sci. USA92, 1287.
Hamada, W., et al. 1994. Curr. Genetics 26, 251.
Hubbard, S. C., et al. 1994. J. Biol. Chem.269, 3717.
Rikkerink, E. H., et al. 1994. Current Genetics25, 202.
Sugimoto, K., et al. 1994. Plant J.5, 863.
Buchschacher, G.L., Jr., and Panganiban, A.T. 1992. J. Virol.66, 2731.
Gaken, J., et al. 1992. Biotechniques13, 32.
Lama, J., and Carrasco, L. 1992. J. Biol. Chem. 267, 15932.
Ma, H., et al. 1992. Gene117, 161.
MacIntyre, G., et al. 1992. Adv. Exp. Med. Biol.276, 67.
Norman, J.A., et al. 1992. Mol. Pharmacol.41, 53.
Bilang, R., et al. 1991. Gene100, 247.
Dale, E., and Ow, D. 1991. Proc. Natl. Acad. Sci. USA88, 10558.
Lee, M.G-S., and Van der Ploeg, L. 1991. Gene105, 255.
Leslie, J.F., and Dickman, M.B. 1991. Applied Environ. Microbiol.57, 1423.
MacIntyre, G., et al. 1991. Antimicrob. Agents Chemother.35, 2125.
Zhou, J., et al. 1991. Gene107, 307.
Bulte, L., and Bennoun, P. 1990. Current Genetics18, 155.
Giordano, T.J., and McAllister, W.T. 1990. Gene88, 285.
Salauze, D., et al. 1990. Antimicrob. Agents Chemother.24, 1915.
Carrasco, L., et al. 1989. Pharmacol. Ther.9, 311.
Crespi, C.L., et al. 1989. Carcinogenesis10, 295.
Damm, B., et al. 1989. Mol. Gen. Genetics217, 6.
Egelhoff, T.T., et al. 1989. Mol. Cell. Biol.9, 1965.
Kronstad, J.W., et al. 1989. Gene79, 97.
Perlin, D.S., et al. 1988. J. Biol. Chem.263, 118.
Cullen, D., et al. 1987. Gene57, 21.
Gonzalez, A., et al. 1978. Biochim. Biophys. Acta521, 459.
Malpartida, F., et al. 1983. Biochem. Biophys. Res. Commun.117, 6.
Rao, S.N., et al. 1983. Antimicrob. Agents Chemother.24, 689.
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Songhai Tian et al.
Cell host & microbe, 27(5), 782-792 (2020-04-18)
The exotoxin TcsL is a major virulence factor in Paeniclostridium (Clostridium) sordellii and responsible for the high lethality rate associated with P. sordellii infection. Here, we present a genome-wide CRISPR-Cas9-mediated screen using a human lung carcinoma cell line and identify semaphorin
Kirstin Kucka et al.
Cell death & disease, 12(4), 360-360 (2021-04-08)
In the early 1990s, it has been described that LTα and LTβ form LTα2β and LTαβ2 heterotrimers, which bind to TNFR1 and LTβR, respectively. Afterwards, the LTαβ2-LTβR system has been intensively studied while the LTα2β-TNFR1 interaction has been ignored to
Timo Löser et al.
Cells, 10(10) (2021-10-24)
Mitochondria are ubiquitous organelles of eukaryotic organisms with a number of essential functions, including synthesis of iron-sulfur clusters, amino acids, lipids, and adenosine triphosphate (ATP). During aging of the fungal aging model Podospora anserina, the inner mitochondrial membrane (IMM) undergoes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service