Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

382110

Sigma-Aldrich

Histone Acetyltransferase Inhibitor II

The Histone Acetyltransferase Inhibitor II, also referenced under CAS 932749-62-7, controls the biological activity of Histone Acetyltransferase. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Histone Acetyltransferase Inhibitor II, 2,6- bis-(3-Bromo-4-hydroxybenzylidene)cyclohexanone, HAT Inhibitor II, p300/CBP Inhibitor II, GCN5 Inhibitor I, 2,6-bis-(3-Bromo-4-hydroxybenzylidene)cyclohexanone, HAT Inhibitor II, p300/CBP Inhibitor II, GCN5 Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16Br2O3
CAS Number:
Molecular Weight:
464.15
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

ethanol: 10 mg/mL
DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Brc1c(ccc(c1)\C=C2/CCC\C(=C/c3cc(c(cc3)O)Br)\C/2=O)O

InChI

1S/C20H16Br2O3/c21-16-10-12(4-6-18(16)23)8-14-2-1-3-15(20(14)25)9-13-5-7-19(24)17(22)11-13/h4-11,23-24H,1-3H2/b14-8+,15-9+

InChI key

YOLKEKNTCBWPSD-VOMDNODZSA-N

General description

A cell-permeable bis-arylidene cyclohexanone compound that acts as a p300/CBP-selective histone acetyltransferase inhibitor (IC50 = 5 µM), while it affects GCN5 and PCAF only at much higher concentrations (30% and 0% inhibition, respectively, at 5 µM inhibitor concentration). Shown to decrease histone H3 acetylation (IC50 ≤ 40 µM) and induce chromatin condensation in HeLa cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month.

Other Notes

Costi, R., et al. 2007. J. Med. Chem.50, 1973.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lingling Meng et al.
Heliyon, 10(6), e27045-e27045 (2024-03-19)
Imbalances between Bcl-2 and caspase-3 are significant evidence of apoptosis, which is considered an influential factor in rapidly occurring neuronal cell death and the decline of neurological function after stroke. Studies have shown that acupuncture can reduce poststroke brain cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service