Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

374087

Sigma-Aldrich

Anti-Heme Oxygenase-1 Mouse mAb (HO-1-1)

liquid, clone HO-1-1, Calbiochem®

Synonym(s):

Anti-HO-1, Anti-Hsp32

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

HO-1-1, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, rat, canine, monkey, mouse, bovine

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

bovine ... Hmox1(513221)
dog ... Hmox1(442987)
human ... HMOX1(3162)
mouse ... Hmox1(15368)
rat ... Hmox1(24451)
rhesus monkey ... Hmox1(719266)

General description

Anti-Heme Oxygenase-1, mouse monoclonal, clone HO-1-1, recognizes the ~32 kDa HO-1 in a variety of species. It is validated for use in Western blotting, immunocytochemistry, and immunoprecipitation.
Protein G purified mouse monoclonal antibody. Recognizes the ~32 kDa heme oxygenase (HO-1) protein.
Recognizes the ~32 kDa HO-1 protein.

Immunogen

Human
a synthetic peptide (MERPQPDSMPQDLSEALKEATKEVHTQAEN) corresponding to amino acids 1-30 of human HO-1 prepared as a four-branched multiple antigen peptide

Application

Immunoblotting (4 µg/ml, chemiluminescence)

Immunocytochemistry (1:1000)

Immunoprecipitation (20 µg/ml)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 50% glycerol.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Does not cross-react with HO-2. Variables associated with assay conditions will dictate the proper working dilution.
Yoshida, T., et al. 1988. Eur. J. Biochem. 171, 457.
Maines, M.D. 1988. FASEB J.2, 2557.
Trakshel, G.M., et al. 1986 J. Biol. Chem.261, 11131.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hai Lu et al.
Brain sciences, 14(5) (2024-05-25)
The discovery of novel diagnostic methods and therapies for Alzheimer's disease (AD) faces significant challenges. Previous research has shed light on the neuroprotective properties of Apelin-13 in neurodegenerative disorders. However, elucidating the mechanism underlying its efficacy in combating AD-related nerve
László Potor et al.
Oxidative medicine and cellular longevity, 2013, 676425-676425 (2013-06-15)
Oxidized cell-free hemoglobin (Hb), including covalently cross-linked Hb multimers, is present in advanced atherosclerotic lesions. Oxidation of Hb produces methemoglobin (Fe(3+)) and ferryl hemoglobin (Fe(4+) = O(2-)). Ferryl iron is unstable and can return to the Fe(3+) state by reacting
László Potor et al.
Oxidative medicine and cellular longevity, 2018, 3812568-3812568 (2018-03-22)
The infiltration of red blood cells into atheromatous plaques is implicated in atherogenesis. Inside the lesion, hemoglobin (Hb) is oxidized to ferri- and ferrylHb which exhibit prooxidant and proinflammatory activities. Cystathione gamma-lyase- (CSE-) derived H2S has been suggested to possess
Qian Sun et al.
Experimental and therapeutic medicine, 21(3), 190-190 (2021-01-26)
The nuclear erythroid 2-related factor 2 (NRF2)/antioxidant response element (ARE) pathway has been shown to provide strong protection against oxidative stress injury induced by renal ischemia-reperfusion (IR). However, the endogenous regulatory mechanism of the NRF2/ARE pathway in renal IR injury
Steven J T Jackson et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 60, 431-438 (2013-08-14)
Curcumin, a component of turmeric spice that imparts flavor and color to curry, is thought to possess anti-inflammatory and antioxidant properties in biological tissues. However, while such efficacies have been described in the context of carcinogenesis, the impact of curcumin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service