Skip to Content
MilliporeSigma

370656

Guanosine 3′,5′-cyclic Monophosphate, Sodium Salt

Important intracellular second messenger implicated in the action of nitric oxide in many cell types.

Synonym(s):

Guanosine 3′,5′-cyclic Monophosphate, Sodium Salt, Cyclic GMP, G-3,5-MP, cGMP, Na

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H11N5O7P · Na
CAS Number:
Molecular Weight:
367.19
UNSPSC Code:
41106305
NACRES:
NA.77
MDL number:
Assay:
>98% (HPLC)
Form:
solid
Quality level:
Storage condition:
OK to freeze, desiccated (hygroscopic)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

description

Merck USA index - 14, 2709

assay

>98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].[P]1(=O)(O[C@@H]2[C@H](O[C@H]([C@@H]2O)[n]3c4nc(nc(c4nc3)[O-])N)CO1)O

InChI

1S/C10H12N5O7P.Na/c11-10-13-7-4(8(17)14-10)12-2-15(7)9-5(16)6-3(21-9)1-20-23(18,19)22-6;/h2-3,5-6,9,16H,1H2,(H,18,19)(H3,11,13,14,17);/q;+1/p-1/t3-,5-,6-,9-;/m1./s1

InChI key

KMPIYXNEROUNOG-GWTDSMLYSA-M

General description

An important intracellular second messenger implicated in mediating the action of nitric oxide in many cell types. Found to activate protein kinase G. Suppresses apoptosis in rat ovarian follicles.
Important intracellular second messenger implicated in the action of nitric oxide in many cell types. Activates protein kinase G. Suppresses apoptosis in rat ovarian follicles.

Biochem/physiol Actions

Primary Target
Activates protein kinase G
Product does not compete with ATP.
Reversible: no
Cell permeable: no

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 4 months at -20°C.

Other Notes

Chun, S.Y., et al. 1995. Endocrinology136, 3120.
Kaupp, U.B., and Koch, K.W. 1992. Annu. Rev. Physiol.54, 153.
Goldberg, N.D., and Haddox, M.K. 1977. Annu. Rev. Biochem. 46, 823.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
370656-50MG04055977191660