Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

361549

Sigma-Aldrich

GSK-3β Inhibitor VIII

The GSK-3β Inhibitor VIII, also referenced under CAS 487021-52-3, controls the biological activity of GSK-3β. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

GSK-3β Inhibitor VIII, AR-A014418, N-(4-Methoxybenzyl)-Nʹ-(5-nitro-1,3-thiazol-2-yl)urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N4O4S
CAS Number:
Molecular Weight:
308.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[s]1c(ncc1[N+](=O)[O-])NC(=O)NCc2ccc(cc2)OC

InChI

1S/C12H12N4O4S/c1-20-9-4-2-8(3-5-9)6-13-11(17)15-12-14-7-10(21-12)16(18)19/h2-5,7H,6H2,1H3,(H2,13,14,15,17)

InChI key

YAEMHJKFIIIULI-UHFFFAOYSA-N

General description

A cell-permeable thiazole-containing urea compound that acts as a potent inhibitor of GSK-3. Shown to inhibit GSK-3β with high potency (IC50 = 104 nM). Inhibition is competitive with respect to ATP (Ki = 38 nM). Specificity has been reported using a panel of 28 different kinases, including Cdk2 and Cdk5 (IC50 >100 µM). Shown to prevent tau phosphorylation at a GSK-3-specific site. Also shown to protect neuronal cells against Aβ-mediated neurodegeneration in vitro and reduce tauopathy in mice in vivo (30 µmol/kg delivered with 40% PEG400 and 40% dimethylacetamide in water). Caution: Do not use delivering vehicles containing dimethylamine for in vivo studies, as it is highly toxic to animals. A 25 mM (5 mg/649 µl) solution of GSK-3β Inhibitor VIII (Cat. No. 361557) in DMSO is also available.
A cell-permeable, potent inhibitor of glycogen synthase kinase-3 (GSK-3). Shown to inhibit GSK-3β with high potency (IC50 = 104 nM). Inhibition is competitive with respect to ATP (Ki = 38 nM). Specificity has been reported using a panel of 28 different kinases, including Cdk2 and Cdk5 (IC50 >100 µM). Shown to prevent tau phosphorylation at a GSK-3-specific site. Also shown to protect neuronal cells against Aβ-mediated neurodegeneration in vitro and reduce tauopathy in mice in vivo (30 µmol/kg delivered with 40% PEG400 and 40% dimethylacetamide in water). Caution: Do not use delivering vehicles containing dimethylamine for in vivo studies, as it is highly toxic to animals.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Gsk-3β
Product competes with ATP.
Reversible: no
Target IC50: 104 nM against GSK-3β

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Noble, W., et al. 2005. Proc. Natl. Acad. Sci. USA102, 6990.
Bhat, R., et al. 2003. J. Biol. Chem.278, 45937.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suresh L Mehta et al.
Neuromolecular medicine (2022-11-30)
Post-stroke secondary brain damage is significantly influenced by the induction and accumulation of α-Synuclein (α-Syn). α-Syn-positive inclusions are often present in tauopathies and elevated tau levels and phosphorylation promotes neurodegeneration. Glycogen synthase kinase 3β (GSK-3β) is a known promoter of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service