Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

361540

Sigma-Aldrich

GSK-3β Inhibitor I

The GSK-3β Inhibitor I, also referenced under CAS 327036-89-5, controls the biological activity of GSK-3β. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

GSK-3β Inhibitor I, TDZD-8, 4-Benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2O2S
CAS Number:
Molecular Weight:
222.26
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C10H10N2O2S/c1-11-9(13)12(10(14)15-11)7-8-5-3-2-4-6-8/h2-6H,7H2,1H3

InChI key

JDSJDASOXWCHPN-UHFFFAOYSA-N

General description

A cell-permeable, thiadiazolidinone (TDZD) analog that acts as a highly selective, non-ATP-competitive inhibitor of GSK-3β (IC50 = 2 µM). Inhibits Flt-3 and PKC activities (IC50 = 673 nM and 1.4-5.5 µM, respectively). Does not significantly affect the activities of Cdk1/cyclin B, CK-II, PKA, or PKC (IC50 >100 µM). Shown to inhibit collagen- and thrombin-induced platelet aggregation.
A thiadiazolidinone (TDZD) analog that acts as a highly selective, non-ATP competitive inhibitor of GSK-3β (IC50 = 2 µM). Inhibits Flt-3 and PKC activities (IC50 = 673 nM and 1.4-5.5 µM, respectively). Does not significantly affect the activities of Cdk-1/cyclin B, CK-II, and PKA, (IC50 >100 µM). Binds to the active site of GSK-3β.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Gsk-3β
Product does not compete with ATP.
Reversible: no
Target IC50: 2 µM, 673 nM, 1.4-5.5 µM, against GSK-3β, Flt-3, and PKC activities, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Guzman, M.L., et al. 2007. Blood110, 4436.
Barry, F.A., et al. 2003. FEBS Lett.553, 173.
Martinez, A., et al. 2002. J. Med. Chem.45, 1292.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katryna Pampuscenko et al.
Journal of neurochemistry, 154(3), 316-329 (2019-12-14)
Tau is a microtubule-associated protein, found at high levels in neurons, and its aggregation is associated with neurodegeneration. Recently, it was found that tau can be actively secreted from neurons, but the effects of extracellular tau on neuronal viability are unclear.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service