Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

35675

Sigma-Aldrich

β-Glycerophosphate, Disodium Salt, Pentahydrate

A phosphate group donor in matrix mineralization studies that acts as a protein phosphatase inhibitor.

Synonym(s):

β-Glycerophosphate, Disodium Salt, Pentahydrate, β-GP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7Na2O6P · 5H2O
CAS Number:
Molecular Weight:
306.11
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.77

Quality Level

description

Merck USA index - 14, 4486

Assay

≥97% (titration)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

water: 5 mg/mL

anion traces

chloride (Cl-): ≤0.010%
phosphate (PO43-): ≤0.010%

cation traces

As: ≤0.00050%
heavy metals (as Pb): ≤0.0020%

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[P](=O)([O-])([O-])OC(CO)CO.O.O.O.O.O

InChI

1S/C3H9O6P.2Na.5H2O/c4-1-3(2-5)9-10(6,7)8;;;;;;;/h3-5H,1-2H2,(H2,6,7,8);;;5*1H2/q;2*+1;;;;;/p-2

InChI key

PEMUISUYOHQFQH-UHFFFAOYSA-L

Related Categories

General description

A phosphate group donor in matrix mineralization studies. Accelerates calcification in vascular smooth muscle cells. Also acts as a protein phosphatase inhibitor.

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Other Notes

Fujimoto, H., and Mabuchi, I. 1997. J. Biochem. 122, 518.
Lecanda, F., et al. 1997. J. Cell Biochem. 67, 386.
Laurencin, C.T., et al. 1996. Bone 19, 93s.
Shioi, A., et al. 1995. Arterioscler. Thromb. Vasc. Biol. 15, 2003.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonella Stanzione et al.
Biomaterials science, 9(22), 7492-7503 (2021-10-14)
Motor neuron diseases are neurodegenerative diseases that predominantly affect the neuromuscular system. To date, there are no valid therapeutic treatments for such diseases, and the classical experimental models fail in faithfully reproducing the pathological mechanisms behind them. In this regard
G Ruiz-Heiland et al.
Clinical oral investigations, 25(4), 1933-1944 (2020-08-21)
Juvenile obesity is a complex clinical condition that is present more and more frequently in the daily orthodontic practice. Over-weighted patients have an impaired bone metabolism, due in part to their increased levels of circulating adipokines. Particularly, leptin has been
Justyna M Kowal et al.
Stem cells translational medicine, 9(2), 189-202 (2019-11-24)
Cultured human bone marrow stromal (mesenchymal) stem cells (hBM-MSCs) are heterogenous cell populations exhibiting variable biological properties. Quantitative high-content imaging technology allows identification of morphological markers at a single cell resolution that are determinant for cellular functions. We determined the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service