Skip to Content
MilliporeSigma

356310

Glyburide

A sulfonylurea that selectively blocks ATP-sensitive K+ channels.

Synonym(s):

Glyburide, Glybenclamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H28ClN3O5S
CAS Number:
Molecular Weight:
494.00
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (TLC)
Form:
solid
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

Merck USA index - 14, 4478

assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 25 mg/mL, ethanol: 5 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

[S](=O)(=O)(NC(=O)NC3CCCCC3)c1ccc(cc1)CCNC(=O)c2c(ccc(c2)Cl)OC

InChI

1S/C23H28ClN3O5S/c1-32-21-12-9-17(24)15-20(21)22(28)25-14-13-16-7-10-19(11-8-16)33(30,31)27-23(29)26-18-5-3-2-4-6-18/h7-12,15,18H,2-6,13-14H2,1H3,(H,25,28)(H2,26,27,29)

InChI key

ZNNLBTZKUZBEKO-UHFFFAOYSA-N

General description

A sulfonylurea that selectively blocks ATP-sensitive K+ channels. Reduced K+ conductance causes membrane depolarization and influx of Ca2+ through voltage sensitive Ca2+ channels. Blocks vascular smooth muscle relaxation produced by KATP channel openers. Inhibits the vasorelaxing effects of nitric oxide.
Antidiabetic sulfonylurea that selectively blocks ATP-sensitive K+ channels. Reduced K+ conductance causes membrane depolarization and influx of Ca2+ through voltage sensitive Ca2+ channels. Blocks vascular smooth muscle relaxation produced by KATP channel openers. High-affinity binding sites are widely distributed and have been identified in the cardiovascular system, the central nervous system, and pancreatic β-cells. In cultured cells, glyburide enhances insulin action and stimulates the synthesis of glucose transporters. Displays large potency difference in action on pancreatic tissue (IC50 in nM range) and vascular tissue (IC50 in µM range).

Biochem/physiol Actions

Cell permeable: no
Primary Target
ATP-sensitive K+ channels
Product does not compete with ATP.
Reversible: no

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Holzmann, S., et al. 1992. Eur. J. Pharmacol.215, 1.
Rabuazzo, A.M., et al. 1992. Endocrinology131, 1815.
Miller, J.A., et al. 1991. J. Pharmacol. Exp. Ther.256, 358.
Jacobs, D.R., et al. 1989. Diabetes 38, 205.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under Glyburide


hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library