Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

35044BC

Millipore

VIP® Gold Listeria Positive Control

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129

shelf life

22 mo.

Quality Level

technique(s)

lateral flow assay: suitable

application(s)

environmental
food and beverages

storage temp.

2-8°C

General description

VIP® Gold Listeria Positive Control, 5mL

Components

One bottle, 5.0ml

Compatibility

BioControl, For use with VIP® Gold

Testing Method

Lateral Flow

Legal Information

VIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service