Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

345670

Sigma-Aldrich

Galanthamine, Hydrobromide

A competitive and reversible inhibitor of acetylcholinesterase.

Synonym(s):

Galanthamine, Hydrobromide, Nivalin, HBr

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO3 · xHBr
CAS Number:
Molecular Weight:
287.35 (free base basis)
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.77

Quality Level

description

Merck USA index - 14, 4340

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

ethanol: 1 mg/mL
water: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Br-].[N+H]1(CC[C@@]32[C@@H](Oc4c3c(ccc4OC)C1)C[C@H](C=C2)O)C

InChI

1S/C17H21NO3.BrH/c1-18-8-7-17-6-5-12(19)9-14(17)21-16-13(20-2)4-3-11(10-18)15(16)17;/h3-6,12,14,19H,7-10H2,1-2H3;1H/t12-,14-,17-;/m0./s1

InChI key

QORVDGQLPPAFRS-XPSHAMGMSA-N

General description

A competitive and reversible inhibitor of acetylcholinesterase. Antimyasthenic agent. Can partially reverse the effects of scopolamine-induced amnesia in rats. Reported to improve learning and short-term memory in animal models.
Cholinesterase antagonist. Antimyasthenic agent. Can partially reverse the effects of scopolamine-induced amnesia in rats.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Acetylcholinesterase
Product competes with ATP.
Reversible: yes

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Pilger, C., et al. 2001. J. Mol. Graph. Model19, 288.
Iliev, A., et al. 1999. Methods Find. Exp. Clin. Pharmacol. 21, 297.
Southan, G.J., et al. 1996. Br. J. Pharmacol.117, 619.
Mutafova-Yambolieva, V.N., et al. 1993. Gen. Pharmacol. 24, 1253.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service