Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

344342

Millipore

D-Fructose-6-phosphate, Disodium Salt

"A glycolytic intermediate formed by the isomerization of glucose-6-phosphate.

Synonym(s):

D-Fructose-6-phosphate, Disodium Salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79

Quality Level

description

Merck USA index - 14, 4276

Assay

≥95% (chromatography)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

impurities

≤0.50% Fructose-1,6-diphosphate

color

white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[P](=O)([O-])([O-])OCC(=O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H13O9P.2Na/c7-1-3(8)5(10)6(11)4(9)2-15-16(12,13)14;;/h3,5-8,10-11H,1-2H2,(H2,12,13,14);;/q;2*+1/p-2/t3-,5-,6-;;/m1../s1

InChI key

ZWRAEINUXILZBD-ABICQQBESA-L

General description

A glycolytic intermediate formed by the isomerization of glucose-6-phosphate.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service