Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

342500

Sigma-Aldrich

FK-506, Streptomyces sp.

Immunosuppressant that blocks T cell proliferation in vitro by inhibiting the generation of several lymphokines, especially IL-2.

Synonym(s):

FK-506, Streptomyces sp., Tacrolimus

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H69NO12
CAS Number:
Molecular Weight:
804.02
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

description

RTECS - KD4200000

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

off-white

solubility

DMSO: 10 mg/mL
methanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N21[C@@H](CCCC2)C(=O)O[C@@H]([C@@H]([C@H](CC(=O)[C@@H](\C=C(\C[C@@H](C[C@@H]([C@H]4O[C@]([C@@H](C[C@@H]4OC)C)(C(=O)C1=O)O)OC)C)/C)CC=C)O)C)\C(=C\C3C[C@H]([C@@H](CC3)O)OC)\C.O

InChI

1S/C44H69NO12.H2O/c1-10-13-31-19-25(2)18-26(3)20-37(54-8)40-38(55-9)22-28(5)44(52,57-40)41(49)42(50)45-17-12-11-14-32(45)43(51)56-39(29(6)34(47)24-35(31)48)27(4)21-30-15-16-33(46)36(23-30)53-7;/h10,19,21,26,28-34,36-40,46-47,52H,1,11-18,20,22-24H2,2-9H3;1H2/b25-19+,27-21+;/t26-,28+,29+,30?,31+,32-,33+,34-,36+,37-,38-,39+,40+,44+;/m0./s1

InChI key

NWJQLQGQZSIBAF-HZXKVGIDSA-N

General description

Immunosuppressant that blocks T cell proliferation in vitro by inhibiting the generation of several lymphokines, especially IL-2. Shown to inhibit the activity of FK-506 binding protein, thereby reversing its effects on sarcoplasmic reticulum Ca2+ release. Inhibits the Ca2+-dependent phosphatase, Protein Phosphatase 2B (Calcineurin; Cat. No. 539568), as well as Na+,K+-ATPase in nephron segments. Also shown to inhibit aldosterone-induced synthesis of Giα-3 protein.
Immunosuppressant that blocks T-cell proliferation in vitro by inhibiting the generation of several lymphokines, especially IL-2. Shown to inhibit the activity of FK-506 Binding Protein, thereby reversing its effects on sarcoplamic reticulum Ca2+ release. Shown to inhibit the Ca2+-dependent phosphatase, Calcineurin (Cat. No. 539565), as well as Na+-K+-ATPase in nephron segments. Also shown to inhibit aldosterone-induced synthesis of Giα-3 protein.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Immunosuppressant that blocks T cell proliferation
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Asano, K., et al. 1996. In Vivo 10, 537.
McCall, E., et al. 1996. Circ. Res. 79, 1110.
Raufman, J.P., et al. 1996. J. Biol. Chem. 271, 19877.
Rokaw, M.D., et al. 1996. Am. J. Physiol. 271, C194.
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Repr. 2 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yamini Yadav et al.
Cell communication and signaling : CCS, 21(1), 82-82 (2023-04-22)
PP1γ is one of the isoforms of catalytic subunit of a Ser/Thr phosphatase PP1. The role of PP1γ in cellular regulation is largely unknown. The present study investigated the role of PP1γ in regulating neuronal insulin signaling and insulin resistance
Mohamed Mahameed et al.
Nature communications, 13(1), 7350-7350 (2022-11-30)
Synthetic receptors targeted to the secretory pathway often fail to exhibit the expected activity due to post-translational modifications (PTMs) and/or improper folding. Here, we engineered synthetic receptors that reside in the cytoplasm, inside the endoplasmic reticulum (ER), or on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service