Skip to Content
MilliporeSigma
All Photos(1)

Documents

341506

Sigma-Aldrich

Fetuin, Fetal Bovine Serum

Synonym(s):

Fetuin, Fetal Bovine Serum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.71

biological source

bovine serum

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

technique(s)

cell culture | mammalian: suitable

color

white to light brown

solubility

water: 50 mg/mL

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

Native fetuin from fetal bovine serum. α1-Globulin present in amounts of 40-50% of total serum protein. After birth it is reduced to 40 mg/100 ml. Important supplement to many serum-free media. Inhibits trypsin activity and promotes cell attachment, growth, and differentiation in many different culture systems. Also contains both N- and O-glycosidically linked carbohydrates. Functions as an opsonin for cationic-deactivating molecules in macrophages. Product must not come in contact with ruminating animals or swine.
Native fetuin from fetal bovine serum. An α-globulin that inhibits trypsin activity and promotes cell attachment, growth, and differentiation in many different culture systems. Also contains both N- and O-glycosidically-linked carbohydrates. Functions as an opsonin for cationic-deactivating molecules in macrophages.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long-term storage or refrigerate (4°C) for short-term storage. Stock solutions are stable for up to 6 months at -20°C or for up to 1 week at 4°C.

Other Notes

Wang, H., et al. 1998. Proc. Natl. Acad. Sci. USA 95, 14429.
Nie, Z. 1992. Am. J. Physiol. 263, C551.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service