Skip to Content
MilliporeSigma

33-047

PI 3-Kinase 3-step HTRF® Assay

The PI 3-Kinase 3-Step HTRF Assay is a high-performance assay kit that provides a universal method for testing all Class I PI3-Kinases in a homogeneous format.

Synonym(s):

3-Step HTRF Assay, PI 3-Kinase Assay

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84
eCl@ss:
32161000
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

species reactivity

human, mouse

manufacturer/tradename

HTRF®, Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

detection method

fluorometric (HTRF)

shipped in

dry ice

Gene Information

human ... PIK3CA(5290)

General description

Phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit α (PIK3CA) gene is mapped to human chromosome 3q26.32. This gene codes for the catalytic subunit p110α of class IA PI3-kinase. PI3K signaling pathways regulate various cellular activities and properties including proliferation, survival, apoptosis, motility, and morphology. Mutation or overexpression of the PIK3CA gene is implicated in the development of various human solid tumors including head and neck cancers or pharyngeal cancer and ovarian cancer.

Application

PI 3-Kinase 3-step HTRF® Assay has been used to determine the in vitro inhibitory potencies of phosphatidylinositol 3-kinase (PI3K) δ inhibitors such as MSD-496486311, MSD126796721, idelalisib, and duvelisib across PI3K α, β, γ, and δ isoforms.

Biochem/physiol Actions

Target Sub-Family: PI3K/PI4K

Packaging

Kit capacity: 10,000 wells

Preparation Note

12 months at -70°C

Other Notes

4X Reaction Buffer

PIP2 (substrate, 1mM stock)

DM A Reaction Buffer

DM B Reaction Buffer

DM C Reaction Buffer

BPIP3

Stop Reagent
Research Category
Apoptosis & Cancer
Research Sub Category
Oncology

Legal Information

HTRF is a registered trademark of Cis Bio International
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Explore all of our products under PI 3-Kinase 3-step HTRF® Assay


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library