Skip to Content
MilliporeSigma

324801

Epoxomicin

≥95% (HPLC), liquid, proteasome inhibitor, Calbiochem®

Synonym(s):

InSolution Epoxomicin, Synthetic, Proteasome Inhibitor XIV

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H50N4O7
Molecular Weight:
554.72
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
liquid
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Epoxomicin, InSolution, ≥95%

Quality Segment

assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

shipped in

wet ice

storage temp.

−20°C

General description

Cell permeable: No


Product does not compete with ATP.


Reversible: No

Epoxomicin is a highly effective and selective proteasome inhibitor thatexhibits significant anti-inflammatory properties in vivo. This potent antitumor agent specifically targets the proteasome. Epoxomicin exhibits a stronger inhibitory effect on chymotrypsin-like activity compared to lactacystin and the peptide vinyl sulfone (NLVS). This powerful natural producteffectively inhibits nuclear factor kappa B (NF-κB) activation in vitro and signific antly reduces inflammation in the murine ear edema assay. Epoxomicin prevents the degradation of IκB and inhibits the activation of NF-κBDNA-binding activity. It can stabilize p53 levels in cell culture. It is an irreversible inhibitor of chymotrypsin-like (CT-L), trypsin-like (T-L), and peptidylglutamyl peptide hydrolyzing (PGPH) activities of the proteasome. It modifies the proteasomal catalytic subunits LMP-7, MECL1, and Z[MJ2] .

Application

Epoxomicin has been used incell culture for proteasome inhibition experiments. It has also been used incell culture medium to treat MODE-K cells for stimulation to blockproteasome.

Packaging

Packaged under inert gas

Physical form

A 1 mM (50 µg/90 µl) solution of Epoxomicin, Synthetic (Cat. No. 324800) in DMSO.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


Still not finding the right product?

Explore all of our products under Epoxomicin


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

188.6 °F - closed cup - (Dimethylsulfoxide)

flash_point_c

87 °C - closed cup - (Dimethylsulfoxide)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library