Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

324630

Sigma-Aldrich

EHNA, Hydrochloride

The racemic mixture of a cell-permeable adenine derivative that acts as a PDE II-selective phosphodiesterase inhibitor and adenosine deaminase.

Synonym(s):

EHNA, Hydrochloride, erythro-9-(2-Hydroxy-3-nonyl)adenine, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H23N5O · xHCl
CAS Number:
Molecular Weight:
277.37 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]1(c2ncnc(c2nc1)N)[C@H]([C@@H](O)C)CCCCCC

InChI

1S/C14H23N5O/c1-3-4-5-6-7-11(10(2)20)19-9-18-12-13(15)16-8-17-14(12)19/h8-11,20H,3-7H2,1-2H3,(H2,15,16,17)/t10-,11-/m0/s1

InChI key

IOSAAWHGJUZBOG-QWRGUYRKSA-N

General description

The racemic mixture of a cell-permeable adenine derivative that acts as a PDE II-selective phosphodiesterase inhibitor (IC50 = 0.8 and 2 µM against human and procine PDE II, respectively) and adenosine deaminase (Ki = 4 nM), while exhibiting little or much reduced potency toward PDE I/II/IV isozymes (IC50 >100 µM) of both human and porcine species. EHNA at 10 µM concentration is also reported to effectively and reversibly block the differentiation and maintain the pluripotency of hES (human embryonic stem) cell line SA121 in feeder-free cultures without exogenously added FGF for up to 30 passages in a manner independent of its known ADA and PDE inhibitory activities.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PDE 2
Product does not compete with ATP.
Reversible: no
Target IC50: 0.8 and 2 µM against human and procine PDE II, respectively
Target Ki: 4 nM against adenosine deaminase

Warning

Toxicity: Standard Handling (A)

Other Notes

Burton, P., et al. 2010. Biochem. J.in press.
Lorbar, M., et al. 1999. J. Mol. Cell. Cardiol.31, 401.
Verde, I., et al. 1999. Br. J. Pharmacol.127, 65.
Reynolds, A.J., et al. 1996. Brain Res.798, 67.
Mery, P.F., et al. 1995. Mol. Pharmacol.48, 121.
Podzuweit, T., et al. 1995. Cell. Signal.7, 733.
Bessodes, M., et al. 1982. Biochem. Pharmacol.31, 879.
Bouchard, P., et al. 1981. Proc. Natl. Acad. Sci. USA78, 1033.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service