Skip to Content
MilliporeSigma

324121

DNP-Hemocyanin Conjugate, Keyhole Limpet

DNP-Hemocyanin Conjugate, Keyhole Limpet, is useful for production of antibodies specific for DNP and hemocyanin. Contains ≥400 DNP-groups per molecule of protein.

Synonym(s):

DNP-KLH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.25
UNSPSC Code:
12352202
Form:
solid (fluffy)
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid (fluffy)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

yellow

shipped in

ambient

storage temp.

2-8°C

General description

DNP conjugate useful for production of antibodies specific for DNP and hemocyanin. Contains ≥400 DNP-groups per molecule of protein.
One of the primary aspects of the immune response is the interaction of antigens with lymphocytes to induce the formation of antibodies, that in turn makes the antigen harmless. Much of our current understanding of the antibody response to antigens has been derived by using the antibody-hapten model. One known model uses the dinitrophenyl (DNP) group. Immunization of many mammalian species with DNP-protein conjugates results in production of antibodies specific for DNP and the amino acid side chains to which it is attached. DNP conjugate useful for production of antibodies specific for DNP and hemocyanin. Contains 400 DNP-groups per molecule.

Preparation Note

May require gentle heating and stirring for 1-2 h. Do not use phosphate-buffered saline to dissolve the product.

Other Notes

Little, J.R. and Eiser, H.N. 1967. Methods Immunol. Immunochem.1, 128.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library