Skip to Content
MilliporeSigma

324101

DNP-Albumin Conjugate, Bovine

DNP-Albumin Conjugate, Bovine, is useful for production of antibodies specific for DNP and albumin. Contains >30 DNP-groups per molecule protein.

Synonym(s):

Albumin DNP Conjugate, Bovine Albumin Conjugate, DNP-Albumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.25
UNSPSC Code:
12352202
Form:
solid (Dry)
Technique(s):
ELISA: suitable
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid (Dry)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

technique(s)

ELISA: suitable

color

amber

solubility

water: 1-2 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

Dinitrophenyl (DNP) conjugate is useful to produce antibodies specific for DNP and albumin. It contains >30 DNP-groups per molecule protein.

Application

DNP-albumin conjugate, bovine has been used in passive anaphylaxis to study the biological effects of IgE receptor mimetic peptide (PepE) in vivo.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Pellom, M.I., et al. 1994. Brit. J. Pharmacol.112, 1101.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library