Skip to Content

322326

Diphtheria Toxin, Unnicked

from Corynebacterium diphtheriae, Major band under reduced conditions (SDS-PAGE), <15% nicking, solid, protein synthesis inhibitor, Calbiochem®

Synonym(s):

DTx

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77
Form:
solid
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Diphtheria Toxin, Unnicked, Corynebacterium diphtheriae, Diphtheria toxin catalyzes ADP-ribosylation of eukaryotic aminoacyltransferase II (EF2) using NAD as substrate. Activation requires nicking with a protease followed by reduction with DTT.

Quality Segment

description

RTECS - XW5807200

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

soluble (Aqueous buffers or H2O. Please see vial label for lot-specific reconstitution volume.)

shipped in

ambient

storage temp.

2-8°C

General description

Diphtheria Toxin, Unnicked, from Corynebacterium diphtheriae catalyzes ADP-ribosylation of eukaryotic aminoacyltransferase II (EF2) using NAD as substrate, thereby inhibiting protein synthesis. May also induce inter-nucleosomal breakdown. Causes DNA fragmentation and cytolysis in U937 cells. Activation requires nicking with a protease followed by reduction with DTT. Diphtheria toxin, synthesized and excreted as a proenzyme, is composed of a single polypeptide chain of 63 kDa. For its enzymatic activity to be expressed, the toxin must undergo two covalent alterations in structure. First, a mild proteolysis results in the formation of an enzymatically inactive "nicked toxin," which consists of two major fragments (A and B) linked by a disulfide bond. Reduction of the nicked toxin with thiols (DTT) releases the enzymatically active N-terminal A fragment (24 kDa). The C-terminal B fragment (39 kDa) has no apparent enzymatic activity. The B fragment is required for toxicity and is responsible for recognizing and binding of the toxin to cell surface receptors.

Application

Diphtheria Toxin, Unnicked, Corynebacterium diphtheriae has been used for intraperitoneal administration in mice for cell ablation.

Biochem/physiol Actions

Product does not compete with ATP.
Reversible: no
Cell permeable: no
Note: Toxicity may vary by lot of toxin. Each laboratory should determine the optimum dosage for each particular application.
Primary Target
Eukaryotic aminoacyltransferase II (EF2)

Physical form

Lyophilized from sterile 10 mM Tris, 1 mM EDTA, pH 7.5.

Preparation Note

Following reconstitution, aliquot, quickly freeze on dry ice, and freeze (-70°C). Subsequent thawing should be carried out only at room temperature. For assays employing very low concentrations of diphtheria toxin, the use of a carrier protein, such as BSA or HSA, is recommended.
Please see vial label for lot-specific reconstitution volume.

Analysis Note

Major band (under reducing conditions) of ~63 kDa

Other Notes

Diphtheria toxin is purified from Corynebacterium diphtheriae Park Williams strain 8 by a modified method of Pappenheimer, et al. As assessed by disc electrophoresis run at alkaline pH under non-denaturing conditions, this preparation migrates as a major band at 63 kDa, corresponding to the intact toxin. Two faster, more lightly stained bands (24 and 39 kDa), corresponding to A and B fragments, may be observed. Following trypsin treatment in DTT, diphtheria toxin exhibits high activity when assayed for its ability to ADP-ribosylate EF2. The ED50 for CHO cells is determined to be about 0.4 ng/ml.
Diphtheria toxin is purified from Corynebacterium diphtheriae Park Williams strain 8 by a modified method of Pappenheimer, et al. As assessed by disc electrophoresis run at alkaline pH under non-denaturing conditions, this preparation migrates as a major band at 63 kDa, corresponding to the intact toxin. Two faster, more lightly stained bands (24 and 39 kDa), corresponding to A and B fragments, may be observed. Following trypsin treatment in DTT, diphtheria toxin exhibits high activity when assayed for its ability to ADP-ribosylate EF2. The ED50 for CHO cells is determined to be about 0.4 ng/ml.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful (C)


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library