Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

317210

Millipore

DM-NITROPHEN Reagent, Tetrasodium Salt

Caged Ca2+ chelator that undergoes a major and rapid decrease in Ca2+-binding affinity upon photolysis.

Synonym(s):

DM-NITROPHEN Reagent, Tetrasodium Salt, 1-(2-Nitro-4,5-dimethoxyphenyl)-1,2-diaminoethane-N,N,Nʹ,Nʹ-tetraacetic Acid, 4Na, Dimethoxynitrophenamine, 4Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19N3O12 · 4Na
Molecular Weight:
561.32
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

Assay

≥93% (HPLC)

form

solid

reaction suitability

reagent type: chelator

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

orange

solubility

water: 100 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

Caged Ca2+ chelator that undergoes a major and rapid decrease in Ca2+-binding affinity upon photolysis. Can be microinjected or used with permeabilized cells. May also be useful as caged Mg2+ chelator. Kd (Ca2+) = 5 nM; Kd (Mg2+) = 2-5 µM.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Mulkey, R.M., and Zucker, R.S. 1993. J. Physiol. 462, 243.
Zucker, R.S. 1993. Cell Calcium 14, 87.
McCray, J.A., et al. 1992. Biochemistry31, 8856.
Delaney, K.R., and Zucker, R.S. 1990. J. Physiol.426, 473.
Kaplan, J.H., and Ellis-Davies, G.C. 1988. Proc. Natl. Acad. Sci. USA85, 6571.
Morad, M., et al. 1988. Science241, 842.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service