Skip to Content
MilliporeSigma
All Photos(1)

Documents

260913

Millipore

DNase I, Bovine Pancreas

Native DNase I from bovine pancreas.

Synonym(s):

DNase I, Bovine Pancreas, Deoxyribonuclease I

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

biological source

bovine pancreas

form

lyophilized

specific activity

≥60,000 dornase units/mg dry wt

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

solubility

sodium acetate buffer: soluble
water: soluble

shipped in

ambient

storage temp.

−20°C

General description

Native DNase I from bovine pancreas. An endonuclease that preferentially cleaves phosphodiester linkages adjacent to pyrimidine nucleotides to yield 5′-phosphate terminated polynucleotides with a free hydroxyl group at the 3′ position. Acts on single and double stranded DNA. Note: 1 MU = 1,000,000 Dornase units
Note: 32 Dornase units equal ~1 Kunitz unit; 1 MU = 1,000,000 Dornase units.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One Dornase unit is defined as the amount of enzyme that will cause a decrease of 1.0 relative viscosity unit in a solution of highly polymerized DNA from the initial relative viscosity of 4.0 in 10 min at 30°C.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Takeshita, H., et al. 2000. Biochem. Biophys. Res. Commun.269, 481.
Jones, S.J., et al. 1996. J. Mol. Biol.264, 1154.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Du et al.
Cancer letters, 470, 170-180 (2019-11-26)
Cyclophosphamide treatment on a medium-dose, intermittent chemotherapy (MEDIC) schedule activates both innate and adaptive immunity leading to major regression of implanted gliomas. Here, we show that this MEDIC treatment regimen induces tumor cell autonomous type-I interferon signaling, followed by release
Derek Toms et al.
Frontiers in bioengineering and biotechnology, 8, 534-534 (2020-06-26)
We have developed an accessible software tool (receptoR) to predict potentially active signaling pathways in one or more cell type(s) of interest from publicly available transcriptome data. As proof-of-concept, we applied it to mouse photoreceptors, yielding the previously untested hypothesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service