Skip to Content
MilliporeSigma
All Photos(1)

Documents

260585

Sigma-Aldrich

5′-Deoxy-5′-methylthioadenosine

≥98% (HPLC), solid, MTAP substrate, Calbiochem®

Synonym(s):

5′-Deoxy-5′-methylthioadenosine, MTA, MeSAdo

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15N5O3S
CAS Number:
Molecular Weight:
297.33
UNSPSC Code:
12352208
NACRES:
NA.77

product name

5′-Deoxy-5′-methylthioadenosine, A cell-permeable, reversible, and ATP-competitive naturally-occurring co-product of polyamine biosynthesis that acts as an endogenous substrate of methylthioadenosine phosphorylase (MTAP).

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMF: 25 mg/mL
DMSO: soluble

shipped in

ambient

storage temp.

−20°C

InChI

1S/C11H15N5O3S/c1-20-2-5-7(17)8(18)11(19-5)16-4-15-6-9(12)13-3-14-10(6)16/h3-5,7-8,11,17-18H,2H2,1H3,(H2,12,13,14)

InChI key

WUUGFSXJNOTRMR-UHFFFAOYSA-N

General description

A cell-permeable, reversible, and ATP-competitive naturally-occurring co-product of polyamine biosynthesis that acts as an endogenous substrate of methylthioadenosine phosphorylase (MTAP). Potently inhibits the protein carboxymethyltransferase and induces apoptosis in leukemia U937 cells. Also inhibits FGF-2 receptor tyrosine kinase activity as well as the proliferation of human astrocytes and glioma cells.
A naturally occurring co-product of polyamine biosynthesis that acts as an endogenous substrate of methylthioadenosine phosphorylase (MTAP). A cell-permeable, reversible, and ATP-competitive inhibitor of protein carboxymethyltransferase and induces apoptosis in leukemia U937 cells. Also acts as an inhibitor of NGF-induced tyrosine phosphorylation in PC12 cells and blocks FGF-2 receptor tyrosine kinase in astrocytes and glioma cells.

Biochem/physiol Actions

Primary Target
protein carboxymethyltranferase
Secondary Target
FGF-2receptor tyrosine kinase activity

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze at -20°C. Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Lee, S.H., and Cho, Y.D. 1998. Exp. Cell Res.240, 282.
Smalheiser, N.R. 1998. Brain Res. Dev. Brain Res.51, 53.
Joy, A., et al. 1997. Oncogene14, 171.
Riscoe, M.K., et al. 1984. Biochem. Pharmacol.33, 3639.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service