Skip to Content
MilliporeSigma

239760

β-Cyclodextrin

Homogeneous cyclic α1,4-linked D-glucopyranose units in a seven-member ring. Forms clathrates. Suitable for use with dansyl chloride to form water-soluble complexes for fluorescent labeling of proteins.

Synonym(s):

β-Cyclodextrin, Cycloheptaamylose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C42H70O35
CAS Number:
Molecular Weight:
1134.98
UNSPSC Code:
12352201
NACRES:
NA.79
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

Merck USA index - 14, 2718

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

15-25°C

SMILES string

O1C2OC(C(OC3OC(C(OC4OC(C(OC5OC(C(OC6OC(C(OC7OC(C(OC8OC(C1C(C8O)O)CO)C(C7O)O)CO)C(C6O)O)CO)C(C5O)O)CO)C(C4O)O)CO)C(C3O)O)CO)C(C2O)O)CO

InChI

1S/C42H70O35/c43-1-8-29-15(50)22(57)36(64-8)72-30-9(2-44)66-38(24(59)17(30)52)74-32-11(4-46)68-40(26(61)19(32)54)76-34-13(6-48)70-42(28(63)21(34)56)77-35-14(7-49)69-41(27(62)20(35)55)75-33-12(5-47)67-39(25(60)18(33)53)73-31-10(3-45)65-37(71-29)23(58)16(31)51/h8-63H,1-7H2

InChI key

WHGYBXFWUBPSRW-UHFFFAOYSA-N

General description

Homogeneous cyclic α1,4-linked D-glucopyranose units in a seven member ring. Forms clathrates. Suitable for use with dansyl chloride to form water-soluble complexes for fluorescent labeling of proteins.

Application


  • Preparation and evaluation of fenbendazole methyl-β-cyclodextrin inclusion complexes.: Explores the use of β-cyclodextrin in veterinary medicine, specifically how its complexation with fenbendazole improves the drug′s solubility and efficacy, showcasing the application in improving animal health care (Ding Y et al., 2024).

  • Dynamically Cross-Linked Double-Network Hydrogels with Matched Mechanical Properties and Ideal Biocompatibility for Artificial Blood Vessels.: This research highlights the use of β-cyclodextrin in developing biocompatible hydrogels for artificial blood vessels, pointing to its significant potential in biomedical engineering and regenerative medicine (Jia XY et al., 2024).

  • Structural Features of Styrene-Functionalized Cyclodextrin Polymers That Promote the Adsorption of Perfluoroalkyl Acids in Water.: Demonstrates the environmental application of β-cyclodextrin polymers in water purification, specifically in the adsorption and removal of harmful perfluoroalkyl substances (Lin ZW et al., 2024).

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Armspach, D., et al. 1999. in Bioorganic Chemistry: Carbohydrates (Hecht, S.M., ed.) p. 458, Oxford University Press, New York.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under β-Cyclodextrin


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library