Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

2380

Sigma-Aldrich

Creatine Phosphate disodium salt

≥97% (TLC), crystalline, mitocondrial energy storage, Calbiochem®

Synonym(s):

Creatine Phosphate, Disodium Salt, Phosphocreatine, 2Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N3O5P · 2Na
CAS Number:
Molecular Weight:
255.08
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Creatine Phosphate, Disodium Salt, Creatine Phosphate, Disodium Salt, CAS 19333-65-4, is a high-energy phosphate compound that stores energy from mitochondria whenever sufficient amounts of ATP are present.

Quality Level

description

Merck USA index - 14, 7340

Assay

≥97% (TLC)

form

crystalline

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 150 mg/mL

anion traces

phosphate (PO43-): ≤0.5% (Inorganic)

shipped in

ambient

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC(=O)CN(C)\C(=N\[H])\N)O

InChI

1S/C4H10N3O5P/c1-7(4(5)6)2-3(8)12-13(9,10)11/h2H2,1H3,(H3,5,6)(H2,9,10,11)/p-1

InChI key

KPXOWQOQJKGYRN-UHFFFAOYSA-M

General description

Creatine phosphate is a high-energy phosphate compound that stores energy from mitochondria whenever sufficient amounts of ATP are present. When ATP levels are low, energy from phosphocreatine is transferred to ADP molecules, which are quickly converted to ATP. It is often a component of ATP-generating systems.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tokuda, H., et al. 1990. Eur. J. Biochem.192, 583.
Bessman, S.P., and Carpenter, C.L. 1985. Annu. Rev. Biochem.54, 831.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service