Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

234109-M

Millipore

Colcemid

Cell synchronization agent.

Synonym(s):

Colcemid, Demecolcine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25NO5
CAS Number:
Molecular Weight:
371.43
MDL number:
UNSPSC Code:
12352200

description

Merck USA index - 14, 2887

Assay

≥98% (HPLC)

form

crystalline powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

chloroform: 10 mg/mL
ethanol: soluble

storage temp.

10-30°C

SMILES string

N(C1CCc2c(c(c(c(c2)OC)OC)OC)C3=CC=C(C(=O)C=C31)OC)C

InChI

1S/C21H25NO5/c1-22-15-8-6-12-10-18(25-3)20(26-4)21(27-5)19(12)13-7-9-17(24-2)16(23)11-14(13)15/h7,9-11,15,22H,6,8H2,1-5H3

InChI key

NNJPGOLRFBJNIW-UHFFFAOYSA-N

General description

Cell synchronization agent. Induces apoptosis by blocking mitosis in HeLa S3 cells. Depolymerizes microtubules and limits microtubule formation. Appropriate dilutions inactivate the spindle fiber mechanism during metaphase, systematically immobilizing chromosomes at the metaphase plate.

Biochem/physiol Actions

Primary Target
Depolymerizes microtubules and limits microtubule formation.

Warning

Toxicity: Toxic & Carcinogenic / Teratogenic (G)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) under an inert gas. Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Urbani, L., et al. 1995. Exp. Cell Res. 219, 159.
Kanda, R., et al. 1994. J. Radiat. Res. 35, 41.
Sherwood, S.W., et al. 1994. Exp. Cell Res. 215, 373.
Nishiyama, I., and Fuji, T. 1992. Exp. Cell Res.198, 214.
Spoerri, P.E., and Roisen, F.J. 1992. J. Neurosci. Res.31, 494.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service