Skip to Content
MilliporeSigma

233183

Clodronate disodium Salt

≥97% (titration), bone resorption inhibitor, solid

Synonym(s):

Clodronate, Disodium Salt, Clodronate, Disodium, Tetrahydrate, Dichloromethylene-bisphosphonate, Disodium Tetrahydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
CH2Cl2Na2O6P2 · 4H2O
CAS Number:
Molecular Weight:
360.92
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥97% (titration)
Form:
solid
Quality level:
Storage condition:
OK to freeze, desiccated (hygroscopic)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Clodronate, Disodium Salt, A non-amino, chloro-containing bisphosphonate that acts as a potent inhibitor of osteoclast-mediated bone resorption.

Quality Level

description

Merck USA index - 14, 2371

assay

≥97% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[P](=O)([O-])(O)C([P](=O)([O-])O)(Cl)Cl.O.O.O.O

InChI

1S/CH4Cl2O6P2.2Na.4H2O/c2-1(3,10(4,5)6)11(7,8)9;;;;;;/h(H2,4,5,6)(H2,7,8,9);;;4*1H2/q;2*+1;;;;/p-2

InChI key

XWHPUCFOTRBMGS-UHFFFAOYSA-L

General description

A non-amino, chloro-containing bisphosphonate that acts as a potent inhibitor of osteoclast-mediated bone resorption. It is metabolized into a non-hydrolyzable ATP analog in osteoclasts and macrophages, and induces apoptosis by inhibiting mitochondrial function. An efficient metal chelator that inhibits invasion, adhesion, and migration of PC-3 prostate cancer cells (IC50 = 100 nM).
A non-amino, chloro-containing, bisphosphonate that acts as a potent inhibitor of osteoclast-mediated bone resorption. Metabolized to a non-hydrolyzable ATP analog in osteoclasts and macrophages that induces apoptosis by inhibiting mitochondrial function. An efficient metal chelator that inhibits invasion, adhesion and migration of PC-3 prostate cancer cells (IC50 = 100 nM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Osteoclast-mediated bone resorption
Product does not compete with ATP.
Reversible: no
Target IC50: 100 nM inhibiting invasion, adhesion, and migration of PC-3 prostate cancer cells

Other Notes

Lehenkari, P.P., et al. 2002. Mol. Pharmacol.61, 1255.
Virtanen, S.S., et al. 2002. Cancer Res.62, 2708.
Smith, R., et al. 1971. Lancet1, 945.
Fleisch, H., et al. 1969. Nature223, 211.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
233183-10MG04055977217773