Skip to Content
MilliporeSigma
All Photos(1)

Documents

228250

Sigma-Aldrich

Cholesterol Oxidase, Streptomyces sp.

Cholesterol Oxidase, Streptomyces sp. CAS 9028-76-6, is a native cholesterol oxidase that catalyzes the oxidation of cholesterol to cholesterone and hydrogen peroxide.

Synonym(s):

Cholesterol Oxidase, Streptomyces sp., Cholesterol:oxygen oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

Quality Level

form

lyophilized solid

specific activity

≥3 units/mg solid
≥40 U/mg

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

yellow

solubility

water: 20 mg/mL

foreign activity

Catalase ≤1%
glucose oxidase and uricase ≤0.01%

shipped in

ambient

storage temp.

−20°C

General description

Native cholesterol oxidase from Streptomyces species. Catalyzes oxidation of cholesterol to cholesterone and hydrogen peroxide (isoelectric points: 6.1 and 7.3). Suitable for determination of total cholesterol when coupled with cholinesterase and peroxidase. Has an optimal pH of 6.0-8.0.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will oxidize 1.0 µmol of cholesterol per min at 25°C, pH 7.0.

Other Notes

Gadda, G., et al. 1997. Eur. J. Biochem. 250, 369.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service