Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

227016

Sigma-Aldrich

CCR2 Antagonist

≥97% (racemic mixture, HPLC), solid, CCR2 antagonist, Calbiochem®

Synonym(s):

CCR2 Antagonist, 2-((Isopropylaminocarbonyl)amino)-N-(2-((cis-2-((4-(methylthio)benzoyl)amino)cyclohexyl)amino)-2-oxoethyl)-5-(trifluoromethyl)-benzamide, CC Chemokine Receptor 2 Antagonist

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H34F3N5O4S
CAS Number:
Molecular Weight:
593.66
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

CCR2 Antagonist, The CCR2 Antagonist, also referenced under CAS 445479-97-0, controls the biological activity of CCR2.

Quality Level

Assay

≥97% (racemic mixture, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

ethanol: 6 mg/mL
DMSO: 60 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cis-diamidocyclohexyl urea compound that potently competes against MCP-1 binding to CCR2 (IC50 = 5.1 nM) and effectively inhibits MCP-1/CCR2-mediated PBMCs Ca+2 flux and chemotaxis (IC50 = 18 and 1 nM, respectively), while showing much less effect against eotaxin binding to CCR3 (37% inhibition at 10 µM). Mutageneis analysis indicates CCR2 Thr292 is critical in maintaining the high affinity antagonist binding.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

Gentle warming, rapid stirring, and/or sonication may be required for complete solubilization.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Cherney, R.J., et al. 2008. J. Med. Chem.51, 721.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Ling Wu et al.
Hepatology (Baltimore, Md.), 69(6), 2364-2380 (2019-01-21)
To understand the mechanism(s) of age-dependent outcomes of hepatitis B virus (HBV) infection in humans, we previously established an age-related HBV mouse model in which 6-week-old (N6W) C3H/HeN mice exhibited virus tolerance whereas 12-week-old (N12W) counterparts presented virus clearance. By
Siyu He et al.
Investigative ophthalmology & visual science, 64(12), 13-13 (2023-09-08)
This study aims to investigate the potential involvement of spleen-derived monocytes in the repair process following corneal epithelial abrasion. A corneal epithelial abrasion model was established in male C57BL/6J mice, and the dynamic changes of monocyte subpopulations in the injured
Qingqing Wu et al.
Cell reports. Medicine, 4(12), 101334-101334 (2023-12-21)
The specific mechanism of sodium-glucose cotransporter 2 (SGLT2) inhibitor in heart failure (HF) needs to be elucidated. In this study, we use SGLT2-global-knockout (KO) mice to assess the mechanism of SGLT2 inhibitor on HF. Dapagliflozin ameliorates both myocardial infarction (MI)-
R C Cornelison et al.
NPJ precision oncology, 6(1), 54-54 (2022-07-30)
Glioblastoma is an aggressive brain cancer characterized by diffuse infiltration. Infiltrated glioma cells persist in the brain post-resection where they interact with glial cells and experience interstitial fluid flow. We use patient-derived glioma stem cells and human glial cells (i.e.
Giuseppina Augimeri et al.
JCI insight, 8(18) (2023-08-22)
Patients with triple-negative breast cancer remain at risk for metastatic disease despite treatment. The acquisition of chemoresistance is a major cause of tumor relapse and death, but the mechanisms are far from understood. We have demonstrated that breast cancer cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service