Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

219478

Sigma-Aldrich

Cdk4 Inhibitor III

The Cdk4 Inhibitor III, also referenced under CAS 265312-55-8, controls the biological activity of Cdk4. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Cdk4 Inhibitor III, 5-(N-(4-Methylphenyl)amino)-2-methyl-4,7-dioxobenzothiazole, Ryuvidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12N2O2S
CAS Number:
Molecular Weight:
284.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

black-purple

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[s]1c2c(nc1C)C(=O)C(=CC2=O)Nc3ccc(cc3)C

InChI

1S/C15H12N2O2S/c1-8-3-5-10(6-4-8)17-11-7-12(18)15-13(14(11)19)16-9(2)20-15/h3-7,17H,1-2H3

InChI key

HFPLHASLIOXVGS-UHFFFAOYSA-N

General description

A cell-permeable dioxobenzothiazole compound that acts a selective Cdk4 inhibitor (IC50 = 6.0 µM for Cdk4/D1 and >200 µM for Cdk2/A). Exhibit better cytotoxic potential against cancer cells (IC50 = 0.61, 1.08, 0.30, and 1.21 µg/ml against A 549, Col 1, HL-60, and HepG2 tumor cells, respectively) than Cisplatin (Cat. No. 232120).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Cdk4
Product does not compete with ATP.
Reversible: no
Target IC50: 6.0 µM for Cdk4/D1 and >200 µM for Cdk2/A

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ryu, C.K., et al. 2000. Bioorg. Med. Chem. Lett.10, 461.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service