Skip to Content
MilliporeSigma

219458

D(+)Cellobiose

Substrate for β-glucosidase.

Synonym(s):

D(+)Cellobiose, Glcβ1,4Glc, 4-O-β-D-Glucopyranosyl-D-glucose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
UNSPSC Code:
12352201
NACRES:
NA.79
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥98% (HPLC)

form

crystalline powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

15-25°C

SMILES string

O1C(C(C(C(C1CO)O)O)O)OC2C(OC(C(C2O)O)O)CO

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2

InChI key

GUBGYTABKSRVRQ-UHFFFAOYSA-N

General description

Substrate for β-glucosidase.

Other Notes

Pitson, S.M., et al. 1997. Enzyme Microb. Technol. 21, 182.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
219458-25GM04055977218435