Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

219008

Sigma-Aldrich

Catalase, Human Erythrocytes

Catalase from human erythrocytes, CAS 9001-05-2. Composed of four identical subunits of ~60 kDa each.

Synonym(s):

Catalase, Human Erythrocytes

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

specific activity

≥50,000 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

SMILES string

O(CC)C(=O)c1ccc(cc1)O

InChI

1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3

InChI key

NUVBSKCKDOMJSU-UHFFFAOYSA-N

General description

Catalase, along with superoxide dismutase and glutathione peroxidase, controls the levels of oxygen-derived free radicals in mammalian cells. Together these molecules may function as a somatic oxidant defense. Prevents apoptosis in cultured rat ovarian follicles.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will decompose 1.0 µmol H₂O₂ per min at 25°C, pH 7.0.

Physical form

In 50 mM Tris, pH 8.0.

Preparation Note

Prepared from blood that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Tilly, J.L., and Tilly, K.I. 1995. Endocrinology136, 242.
Bonaventura, J., et al. 1972. Arch. Biochem. Biophys.150, 606.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service