Skip to Content
MilliporeSigma

217660

Anti-CD18 Mouse mAb (IB4)

liquid, ≥95% (SDS-PAGE), clone IB4

Synonym(s):

Anti-β2 Integrin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Clone:
IB4, monoclonal
Species reactivity:
human
Application:
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

IB4, monoclonal

assay

≥95% (SDS-PAGE)

form

liquid

contains

0.5 mg/mL gentamicin sulfate as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

dilution

(Flow Cytometry (5-10 µg/mL)
Frozen Sections
Immunoprecipitation
Cell Inhibition Assays )

isotype

IgG2a

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Anti-CD18, mouse monoclonal, clone IB4, recognizes the ~95 kDa β2 integrin subunit. It is validated for FC, IP, IHC on frozen sections, and cell inhibition assays.
Protein A purified mouse monoclonal antibody. Recognizes the ~95 kDa β2 integrin subunit (CD18).
Recognizes the ~95 kDa β2 integrin subunit. Blocks the binding of ICAM-1 and ICAM-3 to LFA-1.

Immunogen

Human
human peripheral blood monocytes

Application

Flow Cytometry (5-10 µg/ml)

Frozen Sections (see comments)

Immunoprecipitation (see comments)

Cell Inhibition Assays (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 150 mM NaCl, 100 mM KCl, 50 mM sodium phosphate, pH 7.5.

Preparation Note

Following initial use, aliquot and freeze (-20°C) for long-term storage. Avoid freeze/thaw cycles.

Analysis Note

Negative Control
Nalm-6 cells
Positive Control
HPB-MLT cells

Other Notes

Berman, M.E., et al. 1996. J. Immunol.156, 1515.
Kaneko, M., et al. 1995. J. Immunol. 155, 2631.
Mazzone, A., and Ricevuti, G. 1995. Haematologica80, 161.
Kaneko, M., et al. 1995. J. Immunol.155, 2631.
Landis, R.C., et al. 1994. J. Cell Biol.126, 529.
Wright, S.D., et al. 1983. Proc. Natl. Acad. Sci. USA80, 5699.
Blocks the binding of ICAM-1 and ICAM-3 to LFA-1. FACS was performed on 5x105 cultured human HPB-MLT cells using anti-CD18 antibody at 5 µg/ml and FITC-conjugated goat anti-mouse IgG. This antibody is also reported to work for immunoprecipitation and frozen sections. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Yuan Han et al.
International journal of biological sciences, 20(6), 2072-2091 (2024-04-15)
Background: It had been shown that selective cardiac vagal activation holds great potential for heart regeneration. Optogenetics has clinical translation potential as a novel means of modulating targeted neurons. This study aimed to investigate whether cardiac vagal activation via optogenetics
Di Chen et al.
JCI insight, 7(4) (2022-01-12)
Capillary malformation-arteriovenous malformation (CM-AVM) is a blood vascular anomaly caused by inherited loss-of-function mutations in RASA1 or EPHB4 genes, which encode p120 Ras GTPase-activating protein (p120 RasGAP/RASA1) and Ephrin receptor B4 (EPHB4). However, whether RASA1 and EPHB4 function in the
Asif J Iqbal et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 36(1), e22065-e22065 (2021-12-01)
Neutrophil trafficking is a key component of the inflammatory response. Here, we have investigated the role of the immunomodulatory lectin Galectin-9 (Gal-9) on neutrophil recruitment. Our data indicate that Gal-9 is upregulated in the inflamed vasculature of RA synovial biopsies



Global Trade Item Number

SKUGTIN
217660-100UG04055977202946