Skip to Content
MilliporeSigma

217386

Carboxy-PTIO Sodium Salt

≥97% (TLC), Nitric oxide scavenger, solid

Synonym(s):

Carboxy-PTIO, Sodium Salt, 2-(4-Carboxyphenyl)-4,4,5,5-tetramethylimidazoline-1-oxyl-3-oxide, Na

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H16N2O4 · Na
Molecular Weight:
299.28
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥97% (TLC)
Form:
solid
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Carboxy-PTIO, Sodium Salt, Reacts with nitric oxide at pH 7.4, resulting in the generation of NO2⁻/NO3⁻ with a rate constant of 1 x 10⁴ M⁻¹sec⁻¹.

Quality Segment

assay

≥97% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

color

dark blue

solubility

HEPES Buffer: 0.4 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Reacts with nitric oxide at pH 7.4, resulting in the generation of NO2-/NO3- with a rate constant of 1 x 104 M-1sec-1. Prevents SIN-1 induced endothelial cell death. Reported to have potent therapeutic value in endotoxin shock through its nitric oxide scavenging action.

Biochem/physiol Actions

Reversible: no
Cell permeable: no
Primary Target
Nitric oxide scavenging action
Product does not compete with ATP.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, refrigerate (4°C). Aqueous stock solutions are stable for up to 1 week at 4°C.

Other Notes

Ishii, M., et al., 1999. J. Cardiovasc. Pharmacol. 33, 295.
Amano, F., and Nova, J. 1995. FEBS Lett.368, 425.
Hogg, N., et al. 1995. Free Radical Res.22, 47.
Az-ma, T., et al. 1994. Life Sci.54, 185.
Yoshida, M., et al. 1994. Biochem. Biophys. Res. Commun.202, 923.
Akaike, T., et al. 1993. Biochemistry32, 827.
Yoshida, K., et al. 1993. Infect. Immun.61, 3552.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library