Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

216100-M

Millipore

Carboplatin

An analog of Cisplatin with reduced nephrotoxicity. Displays potent antitumor activity.

Synonym(s):

Carboplatin, cis-Diamine[1,1-cyclobutanedicarboxylato]platinum(II), JM-8, Paraplatin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2O4Pt
CAS Number:
Molecular Weight:
371.25
MDL number:
UNSPSC Code:
12352200

Quality Level

description

Merck USA index - 14, 1822

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water with 5% glucose: 10 mg/mL

storage temp.

10-30°C

SMILES string

[Pt]1OC(=O)C2(CCC2)C(=O)O1.[N+H4].[N+H4]

InChI

1S/C6H8O4.2H3N.Pt/c7-4(8)6(5(9)10)2-1-3-6;;;/h1-3H2,(H,7,8)(H,9,10);2*1H3;/q;;;+2

InChI key

OLESAACUTLOWQZ-UHFFFAOYSA-N

General description

An analog of Cisplatin (Cat. No. 232120) with reduced nephrotoxicity. Displays potent antitumor activity.


Note: Athough soluble in DMSO, Carboplatin is reported to be rendered inactive due to ligand displacement by the nucleophilic sulfur of DMSO. Water with or without 5% glucose is recommended for solubilization prior to culture treatment.
An analog of cisplatin with reduced nephrotoxicity. Displays potent antitumor activity.



Note: Athough soluble in DMSO, Carboplatin is reported to be rendered inactive due to ligand displacement by the nucleophilic sulfur of DMSO. Water with or without 5% glucose is recommended for solubilization prior to culture treatment.

Biochem/physiol Actions

Primary Target
Potent antitumor activity

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hall, M.D., et al. 2014. Cancer Res. In press.
Smith, I.E., et al. 1985. Cancer Treat. Rep. 69, 43.
Harland, S.J., et al. 1984. Cancer Res. 44, 1693.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Muta. 1B - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service