Skip to Content
MilliporeSigma

205541

Cacodylic Acid, Sodium Salt, Trihydrate

Electron microscopy grade. A carcinogen that also acts as a nephrotoxin. Major metabolite of inorganic arsenics.

Synonym(s):

Cacodylic Acid, Sodium Salt, Trihydrate, Sodium Dimethyl Arsenate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C2H6AsO2 · Na · 3H2O
CAS Number:
Molecular Weight:
214.03
UNSPSC Code:
12161700
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

Merck USA index - 14, 1604

assay

≥98% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic)

color

white

pKa (20 °C)

6.27

solubility

water: 200 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[As](=O)([O-])(C)C.[Na+]

InChI

1S/C2H7AsO2.Na/c1-3(2,4)5;/h1-2H3,(H,4,5);/q;+1/p-1

InChI key

IHQKEDIOMGYHEB-UHFFFAOYSA-M

General description

Electron microscopy grade. A carcinogen and a nephrotoxin. Major metabolite of inorganic arsenics.
Electron microscopy grade. A carcinogen that also acts as a nephrotoxin. Major metabolite of inorganic arsenics.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Yamamoto, S., et al. 1995. Cancer Res. 55, 1271.
Murai, T., et al. 1993. Toxicol. Lett. 66, 53.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Toxic & Carcinogenic / Teratogenic (G)


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library